Skip to Content
Merck
All Photos(1)

Key Documents

616467

Sigma-Aldrich

ZDON

≥98% (HPLC), powder, transglutaminase 2 inhibitor, Calbiochem®

Synonym(s):

Transglutaminase 2 Inhibitor, ZDON, Z-DON, 6-diazo-5-oxo-norleucine tetrapeptide, TG2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$447.00

$447.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
$447.00

About This Item

Empirical Formula (Hill Notation):
C31H45N7O9
Molecular Weight:
659.73
UNSPSC Code:
12352200
NACRES:
NA.54

$447.00


Estimated to ship on29 May 2025


Product Name

Transglutaminase 2 Inhibitor, ZDON, A cell-permeable, peptide-based, irreversible inhibitor of transglutaminase 2 (TG2; IC₅₀ = 150 nM for recombinant TG2). Reacts with the active site cysteine of TG2.

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, peptide-based (Z- QVPL) irreversible inhibitor of transglutaminase 2 (TG2) (IC50 = 150 nM for recombinant TG2) that acts by reacting with the active site cysteine of TG2. Exhibits far less potency against TG1 and TG3. Shown to significantly increase PGC-1a and cytochrome c mRNA levels in Q7 and Q111 cells (~50 mM). Also shown to enhance cytochrome c promoter activity by 250% in Q111 cells. Exhibits protective effect in cells exposed to 3-nitropropionic acid and NMDA, however, it does not directly affect mitochondrial bioenergetics.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

McConoughey, S. J., et al. 2010. EMBO Mol. Medicine.2, 349.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min-Su Han et al.
Scientific reports, 10(1), 13228-13228 (2020-08-09)
Transglutaminase 2 (TG2), also known as tissue transglutaminase, is a calcium-dependent enzyme that has a variety of intracellular and extracellular substrates. TG2 not only increases in osteoarthritis (OA) tissue but also affects the progression of OA. However, it is still
Manuel Lisetto et al.
International journal of molecular sciences, 24(15) (2023-08-12)
Transglutaminase 2 (TG2) is a multifunctional protein widely distributed in various tissues and involved in many physiological and pathological processes. However, its actual role in biological processes is often controversial as TG2 shows different effects in these processes depending on
Anurag Rathore et al.
Scientific reports, 10(1), 5350-5350 (2020-03-27)
The major barrier to a HIV-1 cure is the persistence of latent genomes despite treatment with antiretrovirals. To investigate host factors which promote HIV-1 latency, we conducted a genome-wide functional knockout screen using CRISPR-Cas9 in a HIV-1 latency cell line

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service