Skip to Content
Merck
All Photos(1)

Key Documents

616453

Sigma-Aldrich

TGF-β RI Kinase Inhibitor III

The TGF-β RI Kinase Inhibitor III, also referenced under CAS 356559-13-2, controls the biological activity of TGF-β RI Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

TGF-β RI Kinase Inhibitor III, Transforming Growth Factor-β Type I Receptor Kinase Inhibitor III, 2-(5-Benzo[1,3]dioxol-4-yl-2-tert-butyl-1H-imidazol-4-yl)-6-methylpyridine, HCl, ALK5 Inhibitor III, SB-505124, HCl

Sign Into View Organizational & Contract Pricing

Select a Size

2 MG
$303.00

$303.00


Estimated to ship on22 May 2025



Select a Size

Change View
2 MG
$303.00

About This Item

Empirical Formula (Hill Notation):
C20H21N3O2 · xHCl
CAS Number:
Molecular Weight:
335.40 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$303.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light yellow

solubility

DMSO: 20 mg/mL
water: 3 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Cl.[nH]1c(nc(c1c3cc4c(cc3)OCO4)c2nc(ccc2)C)C(C)(C)C

InChI

1S/C20H21N3O2.ClH/c1-12-6-5-7-14(21-12)18-17(22-19(23-18)20(2,3)4)13-8-9-15-16(10-13)25-11-24-15;/h5-10H,11H2,1-4H3,(H,22,23);1H

InChI key

BTUOOXPZOVNPMF-UHFFFAOYSA-N

General description

A cell-permeable imidazolyl-pyridine compound that acts as a potent, ATP-competitive, reversible, and selective inhibitor of activin receptor-like kinase 4 (IC50 = 129 nM), 5 ( IC50 = 47 nM), and 7. It inhibits p38 MAPKα only at much higher concentrations (IC50 = 10.6 µM) and has little effect against ALK1/2/3/6 as well as a panel of 26 other kinases.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Activin receptor-like kinase 4, 5, 7
Product competes with ATP.
Reversible: yes
Target IC50: 129 nM, 47 nM, against activin receptor-like kinase 4, 5, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hu, T., et al. 2005. Am. J. Physiol. Renal Physiol.289, F816.
DaCosta Byfield, S., et al. 2004. Mol. Pharmacol.65, 744.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service