Skip to Content
Merck
All Photos(1)

Key Documents

612151

Sigma-Aldrich

Tissue Factor, Apo-, Human, Recombinant, S. frugiperda

Synonym(s):

rhTF

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$2,210.00

$2,210.00


Estimated to ship on23 May 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$2,210.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25

$2,210.00


Estimated to ship on23 May 2025


Request a Bulk Order

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

−20°C

General description

M.W. 35,000-37,000.
Recombinant, human apo-tissue factor expressed in S. frugiperda insect cells using a baculovirus expression system. Cell-surface protein of endothelial cells and monocytes involved in coagulation and inflammation. Primary cellular initiator of coagulation protease cascade. Functions as receptor and cofactor for coagulation Factors VII and VIIa. Requires lipidation for clotting assays or other functional activities.

Warning

Toxicity: Standard Handling (A)

Physical form

Solid lyophilized from 200 mM mannitol, 150 mM NaCl, 10 mM Tris-HCl, 0.01% CHAPS, pH 8.0.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C).
Reconstitute with 1 ml distilled H₂O

Other Notes

Wildgoose, P., et al. 1993. Biochemistry 32, 114.
Neuenschwander, PF., et al. 1993. J. Biol. Chem.268, 21489.
Yamamoto, M., et al. 1992. J. Biol. Chem.267, 19089.
Morrissey, J., et al. 1991. Thrombosis Res.52, 247.
Drake, T.A., et al. 1989. J. Cell Biol.109, 389.
Bach, R., et al. 1981. J. Biol. Chem.256, 8324.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service