Skip to Content
Merck
All Photos(1)

Key Documents

610205

Sigma-Aldrich

Anti-Tie2 Mouse mAb (Ab33)

liquid, clone Ab33, Calbiochem®

Synonym(s):

Anti-Angiopoietin Receptor, Anti-Tek

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$510.00

$510.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
$510.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$510.00


Estimated to ship on18 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

Ab33, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, porcine, mouse, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG1

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TEK(7010)

General description

Protein G purified mouse monoclonal antibody. Recognizes the ~140-145 kDa Tie2 protein.
Recognizes the ~140-45 kDa Tie2 protein.
This Anti-Tie2 Mouse mAb (Ab33) is validated for use in Frozen Sections, Immunoblotting, Immunoprecipitation, Paraffin Sections for the detection of Tie2.

Immunogen

Human
a recombinant protein consisting of amino acids 1-745 of human Tie2 fused to a His•Tag sequence

Application

Frozen Sections (see comments)

Immunoblotting (0.2-1 µg/ml, chemiluminescence)

Immunoprecipitation (see comments)

Paraffin Sections (not recommended)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 100 mM Tris-glycine, 30% glycerol, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

For frozen sections, tissues were treated with Triton-X-100 detergent. This antibody has also been reported to work for immunoprecipitation. Variables associated with assay conditions will dictate the proper working dilution.
Lin, P., et al. 1998. Proc. Natl. Acad. Sci. USA95, 8829.
Wong, A.L., et al. 1997. Circ. Res.81, 567.
Peters, K.G., et al. 1998. Br. J. Cancer77, 51.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nattawat Klomjit et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 37(10), 1844-1856 (2022-04-23)
Renal artery stenosis (RAS) is an important cause of chronic kidney disease and secondary hypertension. In animal models, renal ischemia leads to downregulation of growth factor expression and loss of intrarenal microcirculation. However, little is known about the sequelae of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service