Skip to Content
Merck
All Photos(1)

Key Documents

598250

Millipore

THP

A water-soluble, neutral, and odorless disulfide reducing agent that is more stable and effective than DTT.

Synonym(s):

THP, Tris(hydroxypropyl)phosphine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$169.00

$169.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
$169.00

About This Item

Empirical Formula (Hill Notation):
C9H21O3P
CAS Number:
Molecular Weight:
208.24
MDL number:
UNSPSC Code:
12352128
NACRES:
NA.25

$169.00


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥80% (31P-NMR)

form

viscous liquid

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

colorless

solubility

water: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

P(CCCO)(CCCO)CCCO

InChI

1S/C9H21O3P/c10-4-1-7-13(8-2-5-11)9-3-6-12/h10-12H,1-9H2

InChI key

YICAEXQYKBMDNH-UHFFFAOYSA-N

General description

A water-soluble, neutral, and odorless disulfide reducing agent that is more stable and effective than DTT. THP is comparable with TCEP (Cat. No. 580560) as a reducing agent. Its reduction capacity against oxidized glutathione is 0.232 meq/g (Note: TCEP has a reduction capacity of 0.182 meq/g). THP is also more resistant to air oxidation as compared to DTT.

Packaging

Packaged under inert gas

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service