Skip to Content
Merck
All Photos(1)

Key Documents

573103

Sigma-Aldrich

STAT3 Inhibitor VII

The STAT3 Inhibitor VII, also referenced under CAS 1041438-68-9, controls the biological activity of STAT3.

Synonym(s):

STAT3 Inhibitor VII, Ethyl-1-(4-cyano-2,3,5,6-tetrafluorophenyl)-6,7,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$247.00

$247.00


Estimated to ship on26 May 2025



Select a Size

Change View
5 MG
$247.00

About This Item

Empirical Formula (Hill Notation):
C19H7F7N2O3
CAS Number:
Molecular Weight:
444.26
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$247.00


Estimated to ship on26 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to light orange

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable dihydroquinoline compound that potently and specifically inhibits cellular JAK1/JAK2/TYK2/STAT3 activation (IC50 against U266 constitutive STAT3 Y705 phosphorylation =170 nM) as well as the transcription of STAT3 target genes, while exhibiting little effects against pathways involved in Akt/mTOR, Erk1/2, or STAT5 activation. Since the inhibitor does not affect JAK1/2/3 kinase activity, its cellular target(s) appears to be upstream of JAKs.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Xu, J., et al. 2008. J. Med. Chem.51, 4115.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian G Hunt et al.
Cancers, 14(10) (2022-05-29)
Aberrant RON signaling is present in numerous cancers including breast cancer. Evidence suggests that the ligand, hepatocyte growth factor-like (HGFL), is also overexpressed in breast cancer. RON (MST1R) and HGFL (MST1) genes are located on human chromosome 3 and mouse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service