Skip to Content
Merck
All Photos(1)

Key Documents

572636

Sigma-Aldrich

SU6656

InSolution, ≥95%, inhibitor of Src-family protein tyrosine kinases

Synonym(s):

InSolution SU6656

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$210.00

$210.00


Estimated to ship on22 May 2025



Select a Size

Change View
500 μG
$210.00

About This Item

Empirical Formula (Hill Notation):
C19H21N3O3S
Molecular Weight:
371.45
UNSPSC Code:
12352200
NACRES:
NA.77

$210.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

General description

A potent inhibitor of Src-family protein tyrosine kinases. Inhibits Src (IC50 = 280 nM) as well as closely related kinases Fyn (IC50 = 170 nM), Yes (IC50 = 20 nM) and Lyn (IC50 = 130 nM). The inhibition is competitive with respect to ATP. Has only a trivial effect on Lck (IC50 = 6.88 µM). Does not affect the activity of PDGF-β receptor kinase (IC50 >10 µM) and IGF-1 receptor kinase (IC50 gt;20 µM).

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 280 nM inhibiting Src

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 10 mM (500 µg/135 µl) solution of SU6656 (Cat. No. 572635) in DMSO.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service