Skip to Content
Merck
All Photos(2)

Key Documents

567417

Sigma-Aldrich

SOD1 Inhibitor, LCS-1

The SOD1 Inhibitor, LCS-1, also referenced under CAS 41931-13-9, controls the biological activity of SOD1.

Synonym(s):

SOD1 Inhibitor, LCS-1, lung cancer screen 1, 4,5-dichloro-2-m-tolylpyridazin-3(2H)-one, 4,5-dichloro-2-(3-methylphenyl)-3(2H)-pyridazinone, Superoxide dismutase 1 inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$307.00

$307.00


Estimated to ship on23 May 2025



Select a Size

Change View
10 MG
$307.00

About This Item

Empirical Formula (Hill Notation):
C11H8Cl2N2O
CAS Number:
Molecular Weight:
255.10
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

$307.00


Estimated to ship on23 May 2025


Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

SMILES string

ClC1=C(C(=O)N(N=C1)c2cc(ccc2)C)Cl

InChI

1S/C11H8Cl2N2O/c1-7-3-2-4-8(5-7)15-11(16)10(13)9(12)6-14-15/h2-6H,1H3

InChI key

SYUPLLHVMCLXEM-UHFFFAOYSA-N

General description

A 2-phenylpyridazin-3(2H)-one based non-copper chelater that selectively inhibits the activity of Superoxide dismutase 1 (Cu/Zn-SOD) (IC50 = 1.07 µM). Cells over-expressing SOD1 show reduced sensitivity to LCS-1. Shown to preferably inhibit the growth of a variety of human lung adenocarinoma cell lines with EGFR and KRAS mutations (IC50 = 200 nM). Prevents serum-induced activation of the ERK and PI 3-kinase/AKT signaling thereby affecting cell growth and survival. Apparent half-life of this compound is reported to be 6.6 min in human liver microsomes.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Somwar, R., et al. 2011, PNAS in press.
Somwar, R., et al. 2009. J. Biomol. Screen14, 1176

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Min Ling et al.
Frontiers in oncology, 12, 937444-937444 (2022-08-19)
Gliomas are characterized by high morbidity and mortality, and have only slightly increased survival with recent considerable improvements for treatment. An innovative therapeutic strategy had been developed via inducing ROS-dependent cell death by targeting antioxidant proteins. In this study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service