Skip to Content
Merck
All Photos(1)

Key Documents

566323

Sigma-Aldrich

SIRT1/2 Inhibitor IV, Cambinol

The SIRT1/2 Inhibitor IV, Cambinol, also referenced under CAS 14513-15-6, controls the biological activity of SIRT1/2. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

SIRT1/2 Inhibitor IV, Cambinol, 5-(2-Hydroxy-naphthalen-1-ylmethyl)-6-phenyl-2-thioxo-2,3-dihydro-1H-pyrimidin-4-one, NSC-112546, SIRT1 Inhibitor II, SIRT2 Inhibitor VI

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$468.00

$468.00


Estimated to ship on08 April 2025



Select a Size

Change View
5 MG
$468.00

About This Item

Empirical Formula (Hill Notation):
C21H16N2O2S
CAS Number:
Molecular Weight:
360.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$468.00


Estimated to ship on08 April 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

S=[c]1[nH][c](c(c([nH]1)c4ccccc4)Cc2c3c(ccc2O)cccc3)=O

InChI

1S/C21H16N2O2S/c24-18-11-10-13-6-4-5-9-15(13)16(18)12-17-19(14-7-2-1-3-8-14)22-21(26)23-20(17)25/h1-11,24H,12H2,(H2,22,23,25,26)

InChI key

RVNSQVIUFZVNAU-UHFFFAOYSA-N

General description

A cell-permeable β-naphthol compound that inhibits the NAD-dependent deacetylase activity of hSIRT1 and hSIRT2 (IC50 = 56 µM and 59 µM, respectively) in a substrate-, but not NAD-, competitive manner. It inhibits SIRT5 deacetylase activity only at much higher concentrations (IC50 >300 µM) and exhibits no inhibition against class I and II HDACs. Cambinol-induced apoptosis in BCL6-expressing Burkitt lymphoma cell lines has been attributed to the hyperacetylation of BCL6 and p53. Shown to effectively inhibit xenografted Daudi Burkitt lymphoma growth in mice in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
hSIRT1 and hSIRT2
Product does not compete with ATP.
Reversible: no
Target IC50: 56 µM and 59 µM against the NAD-dependent deacetylase activity of hSIRT1 and hSIRT2, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C.

Other Notes

Heltweg, B., et al. 2006. Cancer Res.66, 4368.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service