Skip to Content
Merck
All Photos(1)

Key Documents

565761

Sigma-Aldrich

γ-Secretase Inhibitor IV

The γ-Secretase Inhibitor IV controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.

Synonym(s):

γ-Secretase Inhibitor IV, 2-Naphthoyl-VF-CHO, N-(2-Naphthoyl)-Val-phenylalaninal

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$320.00

$320.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 MG
$320.00

About This Item

Empirical Formula (Hill Notation):
C25H26N2O3
Molecular Weight:
402.49
UNSPSC Code:
12352200
NACRES:
NA.77

$320.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable, reversible inhibitor of γ-secretase. Equipotently inhibits the release of Aβx-40 (ED50 = 2.6 µM) and Aβx-42 (ED50 = 2.7 µM) in HEK293 cells stably transfected with the Amyloid Precursor Protein Swedish mutants.

Biochem/physiol Actions

Cell permeable: yes
ED50 = 2.6 µM for Aβx-40
Primary Target
x-40
Product does not compete with ATP.
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

2-Naphthoyl-Val-Phe-CHO

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Green, K.N., et al. 2002. J. Physiol.541.3, 1013.
Sinha, S. and Lieberburg, I. 1999. Proc. Natl. Acad. Sci. USA96, 11049.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service