Skip to Content
Merck
All Photos(1)

Key Documents

560000

Millipore

Sputolysin® Reagent

Intended for the isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum.

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
$111.00
1 SET
$740.00

$111.00


Available to ship on31 March 2025Details


Request a Bulk Order

Select a Size

Change View
10 ML
$111.00
1 SET
$740.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161703
NACRES:
NA.25

$111.00


Available to ship on31 March 2025Details


Request a Bulk Order

Quality Level

description

RTECS - EK1610000

form

liquid

reaction suitability

reagent type: reductant

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

shipped in

ambient

storage temp.

15-25°C

SMILES string

SC[C@H](O)[C@@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m0/s1

InChI key

VHJLVAABSRFDPM-IMJSIDKUSA-N

General description

A concentrate of dithiothreitol (DTT) in phosphate buffer. Useful for the isolation of pathogenic or saprophytic bacteria, fungi, yeast, and epithelial cells from sputum and other mucoid fluids.
Concentrate of Dithiothreitol (DTT; Cat. No. 233155) in phosphate buffer, pH 6.5-7.5. Intended for the isolation of epithelial cells, pathogenic or saprophytic bacteria, fungi, and yeasts from sputum. Each vial is sufficient to make 100 ml of working solution.
Note: 1 set = 20 x 10 ml.

Warning

Toxicity: Harmful (C)

Physical form

20 vials each containing 10 ml of concentrated DTT OR 1 vial containing 10 ml of concentrated DTT.

Preparation Note

Dilute the contents of one vial to 100 ml with sterile distilled water. Swirl gently to dissolve any crystals. If the diluted material appears cloudy, the solution may be filtered to remove any insoluble particles. The diluted solution is ~6.5 mM dithiothreitol in 100 mM phosphate buffer, pH 7.0.

Reconstitution

Once a vial is opened the unused concentrate should be purged with an inert gas (e.g. N₂) and refrigerated (4°C). Diluted solutions, stored aseptically under an inert gas, are stable for up to 48 h at 4°C.

Other Notes

Hirsch, S.R., et al. 1969. J. Lab. Clin. Med.74, 346.
Shah, R.R. and Dye, W.E. 1966. Am. Rev. Respir. Dis.94, 454.
Dixon, J.M.S. and Miller, D.C. 1965 (November). Lancet p. 1046.
Cleland, W.W. 1964. Biochemistry3, 480.
May, J.R. 1953 (September). Lancet p. 535.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
SPUTOLYSIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

  1. Is there any significant interference in the signal after using 560000 for sample preparation and PCR analysis?

    1 answer
    1. The presence of DTT in 560000 may be inhibiting signal detection by quenching the fluorescence reading from PCR. It is advisable to remove DTT before conducting PCR analysis.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service