Skip to Content
Merck
All Photos(1)

Key Documents

559290

Sigma-Aldrich

S-100b Protein, Bovine Brain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77
Pricing and availability is not currently available.

Assay

≥95% (SDS-PAGE)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

≤10% S100 α-chain (Western blot)

solubility

50 mM Tris buffer, pH 8.0: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Research area: NEUROSCIENCE

Native homodimeric S-100b isoform composed of ββ-subunits, purified from bovine brain. The S100 calcium-binding protein B (S100B) exists in the form of homo- or hetero-dimers comprised of two subunits, A and B. Specifically, the S-100AB and S-100BB types are referred to as S100B protein and are known for their strong association with nervous tissue. This protein can be found in the cytosol of glial and Schwann cells. [1] The gene responsible for encoding the S100B protein is situated in the chromosomal region 21q22. [2] Under nonreducing conditions both the dimeric and monomeric forms may be visible.

Application

S100 calcium-binding protein B (S100B) has been used:
  • to study the interaction between the serotonin 5-HT7 receptor and the calcium-binding protein S100B using surface plasmon resonance-based method. [3]
  • as a probe to detect and study the binding interactions with proteins during the far-western blotting technique. [4]
  • to investigate the role of S100B in relation to blood-brain barrier (BBB) integrity and its potential involvement in the production of autoantibodies against S100B. [5]
  • to study the mechanism of communication between thermogenic adipocytes and sympathetic neurons mediated by Calsyntenin-3β and S100b, providing insights into the regulation of sympathetic innervation in adipose tissue. [6]

Biochem/physiol Actions

The S100 calcium-binding protein B (S100B) is known to interact with the cytoskeleton and plays crucial roles in cell proliferation, survival, regulation of calcium homeostasis, and enzyme activities. S100B has been observed to have several effects, including up-regulating the expression of inducible NOS, inducing the release of nitric oxide (NO) in astrocytes and microglia, up-regulating cyclooxygenase-2 expression in microglia and monocytes, triggering NO-dependent death of astrocytes and neurons, increasing the production of reactive oxygen species in neurons, and causing disruption of lipid homeostasis and cell cycle arrest. Elevated levels of S100B have also been observed in the CSF of patients with brain tumors, as well as in the blood of patients with Alzheimer′s disease and Parkinson′s disease. [2] Elevated serum level of S-100b is used as a marker of stroke and cerebral edema. The S-100 proteins are acidic Ca2+ binding proteins found predominately in glial cells. Evidence indicates that upon binding to Ca2+, S-100 proteins expose a hydrophobic domain to the solvent in a manner similar to calmodulin. Within the cell, S-100 proteins have been reported to regulate adenylate cyclase, aldolase activities, ATPase, microtubules, and protein phosphorylation.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 50 mM Tris-HCl, pH 7.5, 50 mM NaCl, 100 µM DTT.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Momotani, E., et al. 1993. J. Comp. Pathol. 108, 291.
Baudier, J., et al. 1992. Proc. Natl. Acad. Sci. USA 89, 11627.
Donato, R. 1991. Cell Calcium12, 713.
Isobe, T., et al. 1977. Biochim. Biophys. Acta494, 222.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Innervation of thermogenic adipose tissue via a calsyntenin 3?-S100b axis
Zeng X, et al.
Nature, 569(7755), 229-235 (2019)
Erin Bargerstock et al.
PloS one, 9(7), e101477-e101477 (2014-07-06)
S100B is a reporter of blood-brain barrier (BBB) integrity which appears in blood when the BBB is breached. Circulating S100B derives from either extracranial sources or release into circulation by normal fluctuations in BBB integrity or pathologic BBB disruption (BBBD).
Is peripheral immunity regulated by blood-brain barrier permeability changes?
Bargerstock E, et al.
PLoS ONE, 9(7), e101477-e101477 (2014)
Xing Zeng et al.
Nature, 569(7755), 229-235 (2019-05-03)
The sympathetic nervous system drives brown and beige adipocyte thermogenesis through the release of noradrenaline from local axons. However, the molecular basis of higher levels of sympathetic innervation of thermogenic fat, compared to white fat, has remained unknown. Here we
Rüdiger von Bauer et al.
Journal of immunology (Baltimore, Md. : 1950), 191(1), 369-377 (2013-06-05)
Promiscuity of pattern recognition receptors, such as receptor for advanced glycation end products (RAGE), allows for a complex regulatory network controlling inflammation. Scavenging of RAGE ligands by soluble RAGE treatment is effective in reducing delayed-type hypersensitivity (DTH), even in RAGE(-/-)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service