Skip to Content
Merck
All Photos(2)

Key Documents

557352

Sigma-Aldrich

RORα/γ Agonist, SR1078

The RORα/γ Agonist, SR1078 controls the biological activity of RORα/γ. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

RORα/γ Agonist, SR1078, SR1078, N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)-4-(trifluoromethyl)benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$195.00

$195.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$195.00

About This Item

Empirical Formula (Hill Notation):
C17H10F9NO2
CAS Number:
Molecular Weight:
431.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$195.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(NC1=CC=C(C(O)(C(F)(F)F)C(F)(F)F)C=C1)C2=CC=C(C=C2)C(F)(F)F

InChI

1S/C17H10F9NO2/c18-15(19,20)11-3-1-9(2-4-11)13(28)27-12-7-5-10(6-8-12)14(29,16(21,22)23)17(24,25)26/h1-8,29H,(H,27,28)

InChI key

DUXWIYXHHGNUJU-UHFFFAOYSA-N

General description

A cell-permeable diaryl amide derived from T1317 (a known LXR agonist and retinoic acid receptor (ROR) inverse agonist) that acts as a highly selective agonist of RORα/γ and does not exhibit affinity for FXR, LXRa and LXRb. Shown to stabilize p53 in cancer cells and increase the expression of p21 and PUMA. Induces apoptosis in hepatocellular carcinoma (HepG2) cells in a RORα and p53-dependent process. Also reported to increase the expression of Sox4 and REV-ERBα in HepG2 cells. Exhibits suitable pharmacokinetic properties with sustained plasma levels even after 8 hours following a single i.p. injection. Exposure to SR1078 Induces expression of glucose-6-phosphatase and fibroblast growth factor 21 in murine models (10 mg/kg/i.p).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RORα/γ
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, Y., et al. 2010. ACS Chem Biol.5, 1029.
Wang, Y., et al. 2012. PLoS One.7, e34921.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Myrthala Moreno-Smith et al.
Nature communications, 12(1), 4006-4006 (2021-06-30)
MYCN activation is a hallmark of advanced neuroblastoma (NB) and a known master regulator of metabolic reprogramming, favoring NB adaptation to its microenvironment. We found that the expression of the main regulators of the molecular clock loops is profoundly disrupted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service