Skip to Content
Merck
All Photos(1)

Key Documents

556881

Sigma-Aldrich

Ribonuclease Inhibitor, Human, Recombinant, E. coli

The Ribonuclease Inhibitor, Human, Recombinant, E. coli controls the biological activity of Ribonuclease. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

liquid

specific activity

≥20,000 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

foreign activity

Endonuclease and RNase, none detected

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant, human ribonuclease inhibitor expressed in E. coli. Non-competitive inhibitor that inactivates RNase by non-covalent binding. Has been used to improve cDNA synthesis and in vitro RNA synthesis, increase yields of polysomes and aid in the preparation of RNase-free antibodies. Inhibits RNases A, B, and C. Does not inhibit RNase T1 and S1 nuclease from Aspergillus.
Recombinant, human ribonuclease inhibitor expressed in E. coli. Non-competitive inhibitor that inactivates RNase by non-covalent binding. Has been used to improve cDNA synthesis and in vitro RNA synthesis, increase yields of polysomes, and aid in the preparation of RNase-free antibodies. Inhibits RNases A, B, and C. Does not inhibit RNase T1 and S1 nuclease from Aspergillus.

Biochem/physiol Actions

Cell permeable: no
Primary Target
inactivates RNase
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of material that will inhibit 50% of the activity of 5 ng of RNase A.

Physical form

In 50 mM KCl, 20 mM HEPES, 8 mM DTT, 50% glycerol.

Reconstitution

Maintain at -20°C. Avoid freezing solutions. Addition of DTT is recommended to maintain 8 mM level.

Other Notes

Saxena, S.K., et al. 1992. J. Biol. Chem. 267, 21982.
Lee, F.S., et al. 1989. Biochemistry 28, 225.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nida Ali et al.
Nucleic acids research, 48(2), 847-861 (2019-12-06)
RNase E is a 472-kDa homo-tetrameric essential endoribonuclease involved in RNA processing and turnover in Escherichia coli. In its N-terminal half (NTH) is the catalytic active site, as also a substrate 5'-sensor pocket that renders enzyme activity maximal on 5'-monophosphorylated
Vera Thole et al.
BMC genomics, 20(1), 995-995 (2019-12-21)
Flavonoids are produced in all flowering plants in a wide range of tissues including in berry fruits. These compounds are of considerable interest for their biological activities, health benefits and potential pharmacological applications. However, transcriptomic and genomic resources for wild
Asimina Katsiani et al.
The Journal of general virology, 101(6), 676-684 (2020-05-08)
Rose rosette virus (RRV) (genus Emaravirus) is the causal agent of the homonymous disease, the most destructive malady of roses in the USA. Although the importance of the disease is recognized, little sequence information and no full genomes are available
Ihshan Akthar et al.
Reproduction (Cambridge, England) (2019-12-04)
We previously reported that sperm binding to cultured monolayers of bovine uterine epithelial cells induces an acute inflammatory response involving the Toll-like receptor (TLR2) signaling pathway. This response serves to clear the uterus of sperm and thereby prepares the endometrium
Claudia S Barros et al.
STAR protocols, 3(4), 101735-101735 (2022-10-02)
Here, we describe a protocol to remove single identified cells directly from Drosophila living brains and analyze their transcriptome. We detail the steps to harvest fluorescent cells using a capillary under epifluorescence and transmitted light to avoid contamination. We then

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service