Skip to Content
Merck
All Photos(1)

Key Documents

5.55349

Millipore

DiluCult Large Bag Holder

Large Bag Holder, Accessory to hold the sample bag

Synonym(s):

Media Bag Support, Sample bag holder, Stomacher bag holder

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$2,480.00

$2,480.00


Estimated to ship on28 May 2025


Request more information

Select a Size

Change View
1 PKG
$2,480.00

About This Item

UNSPSC Code:
41122409
NACRES:
NA.84

$2,480.00


Estimated to ship on28 May 2025


Request more information

manufacturer/tradename

DiluCult

application(s)

food and beverages
sample preparation

storage temp.

2-30°C

General description

Accurate microbiology results are highly reliant upon the quality of your sample preparation. Streamline your sample enrichment with our DILUCULT gravimetric dilutors. The Large Bag Holder is an accessory used to firmly hold the sample bag for pooling samples thus preventing sample spillage. It is non autoclavable.

Legal Information

DILUCULT is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DiluCult™ gravimetric dilutors automate food sample dilution, enhancing workflow efficiency in sample preparation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service