Skip to Content
Merck
All Photos(1)

Documents

550701-M

Sigma-Aldrich

Pyruvate Oxidase, Aerococcus viridans

Native pyruvate oxidase from Aerococcus viridans. Catalyzes the oxidation of pyruvate to acetylphosphate, carbon dioxide, and hydrogen peroxide in the presence of phosphate.

Synonym(s):

Pyruvate Oxidase, Aerococcus viridans

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

Quality Level

form

lyophilized solid

specific activity

≥25 U/mg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

impurities

≤0.006% GPT

color

yellow

pI 

4.0

solubility

aqueous buffer: soluble

foreign activity

GOT and lactate oxidase ≤0.002%
catalase ≤0.3%

storage temp.

−20°C

General description

Native pyruvate oxidase from Aerococcus viridans. Catalyzes the oxidation of pyruvate to acetylphosphate, carbon dioxide, and hydrogen peroxide in the presence of phosphate.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme which generates 1 µmol of H₂O₂ per min at 37°C, pH 6.5.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hager, L.P. and Lipmann, F. 1955. Methods Enzymol. 1, 482.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service