Skip to Content
Merck
All Photos(2)

Key Documents

550602

Sigma-Aldrich

PKM2 Activator II, DASA

The PKM2 Activator II, DASA modulates the biological activity of PKM2. This small molecule/inhibitor is primarily used for Biochemicals applications.

Synonym(s):

PKM2 Activator II, DASA, Pyruvate Kinase M2 Activator II, 1-(2,6-Difluorophenylsulfonyl)-4-(2,3-dihydrobenzo[b][1,4]dioxin-6-ylsulfonyl)piperazine

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$276.00

$276.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$276.00

About This Item

Empirical Formula (Hill Notation):
C18H18F2N2O6S2
CAS Number:
Molecular Weight:
460.47
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.51

$276.00


Estimated to ship on30 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC(C=CC=C1F)=C1S(N2CCN(S(C3=CC4=C(OCCO4)C=C3)(=O)=O)CC2)(=O)=O

InChI

1S/C18H18F2N2O6S2/c19-14-2-1-3-15(20)18(14)30(25,26)22-8-6-21(7-9-22)29(23,24)13-4-5-16-17(12-13)28-11-10-27-16/h1-5,12H,6-11H2

InChI key

SHWNKRPMUBFWKE-UHFFFAOYSA-N

General description

A cell-permeable diarylsulfonamide (DASA) compound that acts as a potent and selective activator of tumor-specific M2 isozyme of pyruvate kinase (PKM2; AC50 = 65 nM). Pretreatment of A549 (adenocarcinomic human alveolar basal epithelial cells) with DASA-10 (~10 µM) prevents hydrogen peroxide, diamide, and hypoxia-induced inactivation of PKM2. DASA treatment is shown to cause a diminution in cellular glutathione levels, and higher oxidative stress-induced cell death.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Anastasiou, D., et al. 2011. Science334, 1278.
Boxer, M.B., et al. 2010. J. Med. Chem.53, 1048.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elizabeth K Wiese et al.
Nature metabolism, 3(7), 954-968 (2021-07-07)
Pharmacological activation of the glycolytic enzyme PKM2 or expression of the constitutively active PKM1 isoform in cancer cells results in decreased lactate production, a phenomenon known as the PKM2 paradox in the Warburg effect. Here we show that oxaloacetate (OAA)
Kaustav Das Gupta et al.
Cell reports, 30(8), 2712-2728 (2020-02-27)
Histone deacetylases (HDACs) drive innate immune cell-mediated inflammation. Here we identify class IIa HDACs as key molecular links between Toll-like receptor (TLR)-inducible aerobic glycolysis and macrophage inflammatory responses. A proteomic screen identified the glycolytic enzyme pyruvate kinase M isoform 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service