Skip to Content
Merck
All Photos(1)

Key Documents

539779

Millipore

ProteoExtract® Complete Mammalian Proteome Extraction Kit

Synonym(s):

C-PEK Mammalian Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106510
NACRES:
NA.32
Pricing and availability is not currently available.

usage

20 extractions

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

input

sample type: mammalian tissue(s)
sample type: mammalian cultured cells

shipped in

ambient

storage temp.

2-8°C

General description

A complete mammalian proteome extraction kit (C-PEK) designed to extract virtually all proteins from both mammalian tissues and culture cells in a simple two step process within a microcentrifuge tube to minimize protein loss. The extraction results in a total protein sample that is a clarified and non-viscous protein solution showing higher resolution and an increased total number of spots in 2-D gel electrophoresis. Sample size: 1x107 - 1x108 cells or 50-100 mg tissue.

Packaging

1 kit in Glass bottle

Components

Wash Buffer, Resuspension Buffer, Extraction Reagent, Reducing Reagent, Benzonase® Nuclease (Cat. No. 71206), Glass Beads, and a user protocol.

Warning

Toxicity: Multiple Toxicity Values, refer to MSDS

Principle

ProteoExtract® Complete Mammalian Proteome Extraction Kit is designed to extract total proteins from mammalian tissue culture cells or mammalian tissue.

Storage and Stability

  • Wash Buffer
The Wash Buffer is provided ready-to-use. The buffer should be stored at 4°C.
  • Resuspension Buffer
The Resuspension Buffer is provided ready-to-use. It can be stored at 4°C up to 6 month. For prolonged storage freeze the buffer in convenient aliquots at -20°C.

Avoid repeated freezing and thawing!
  • Extraction Reagent
The Extraction Reagent is supplied as dry powder and can be stored unopened at 4°C. The Extraction Reagent should be reconstituted just prior to use as described below. After reconstitution, it can be stored at 4°C up to 6 month. For prolonged storage freeze the buffers in convenient aliquots at -20°C.

Avoid repeated freezing and thawing!
  • Reducing Agent
The Reducing Agent is supplied as dry powder. It can be stored unopened at 4°C. The Reducing Agent should be reconstituted just prior to use as described below. After reconstitution, aliquot the unused material in 60 µl volumes and freeze at -20°C for further use.

Avoid repeated freezing and thawing!
  • Benzonase®
Benzonase® should be stored at 4°C.

Other Notes

Yuan, X., et al. 2002. Electrophoresis 23, 1185. Allen, L. 2000. Functional Genomics Nature 405 819. Herbert, B. 1999. Electrophoresis 20, 660.

Legal Information

Benzonase is a registered trademark of Merck KGaA, Darmstadt, Germany
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
PROTEOEXTRACT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service