Skip to Content
Merck
All Photos(1)

Documents

539745

Sigma-Aldrich

InhibitorSelect 96-Well Protein Kinase Inhibitor Library II - Calbiochem

Sign Into View Organizational & Contract Pricing


About This Item

usage

sufficient for 1 96-well plate(s)

General description

The InhibitorSelect 96-Well Protein Kinase Inhibitor Library II consists of 80, well-characterized, cell permeable, potent and reversible protein kinase inhibitors; the majority of which are ATP-competitive. They are supplied in a convenient 96-well plate format at a concentration of 10 mM in DMSO. The inhibitors in this library will be useful for target identification in drug discovery, biochemical pathway analysis, screening new protein kinases, and other pharmaceutical-related applications.

Application

The InhibitorSelect 96-Well Protein Kinase Inhibitor Library II consists of 80, well-characterized, cell permeable, potent and reversible protein kinase inhibitors.

Physical form

All of the inhibitors are supplied at a concentration of 10 mM in DMSO (50 μl/well).

Reconstitution

The contents of this pouch have been purged under nitrogen and are vacuum sealed. Cut open the pouch, take the plate out of the pouch, and let it thaw at room temperature. Please remove the lid gently. To re-store, purge with an inert gas, replace the lid

Other Notes

Supplied with a CD containing comprehensive documentation for each inhibitor. Please contact technical service for further details.

Signal Word

Warning

Hazard Statements

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service