Skip to Content
Merck
All Photos(1)

Key Documents

539576

Sigma-Aldrich

PKA, Catalytic Subunit, Bovine Heart

Synonym(s):

cAMP-Dependent Protein Kinase Catalytic Subunit, Protein Kinase A, Catalytic Subunit

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$776.00

$776.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
25 μG
$776.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

$776.00


Estimated to ship on01 June 2025


Request a Bulk Order

biological source

bovine heart

Quality Level

Assay

≥90% (SDS-PAGE)

form

liquid

specific activity

≥20,000 units/μg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

technique(s)

activity assay: suitable

suitability

suitable for molecular biology

shipped in

wet ice

storage temp.

−70°C

General description

Research area: Cell Signaling

Native PKA catalytic subunit purified from bovine heart. Protein Kinase A (PKA) is a serine protein kinase that targets serine and threonine residues.[1]

Biochem/physiol Actions

Protein kinases are crucial for integrating signaling networks within eukaryotic cells.[2] It plays an essential role in regulating proliferation and fate determination by influencing Hedgehog signaling transduction.[3] Upon binding of cAMP to the PKA regulatory subunits, the catalytic subunit of PKA dissociates from the PKA holoenzyme and proceeds to phosphorylate target proteins at serine and/or threonine within the recognition motif, Arg-Arg-X-Ser*-X. The purified monomeric catalytic subunit is capable of activation without the need for cAMP and is well-suited for in vitro studies of PKA phosphorylation with purified substrates or when introduced into intact cells or cell extracts.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme required to incorporate 1 pmol phosphate into Kemptide substrate (Cat. No. 05-23-4900) per min at 30°C.

Physical form

In 350 mM K₂HPO₄, 100 µM DTT, pH 6.8.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Corbin, J.D., et al. 1988. Methods Enzymol159, 74.
Rannels, S.R., et al 1983. Methods Enzymol99, 55.
Witt, J.J. and Roskoski, R. Jr. 1975. Anal Biochem.66, 253.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S S Taylor et al.
Biochimica et biophysica acta, 1697(1-2), 259-269 (2004-03-17)
Protein kinases play a critical role in the integration of signaling networks in eukaryotic cells. cAMP-dependent protein kinase (PKA) serves as a prototype for this large and highly diverse enzyme family. The catalytic subunit of PKA provides the best example
Protein Kinase A Activity and Hedgehog Signaling Pathway
Vitamins and Hormones, 88.0, 273-291 (2012)
Protein kinase A
Morrison H
Journal of Separation Science, 173-178 (2021)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service