Skip to Content
Merck
All Photos(1)

Key Documents

5.38769

Sigma-Aldrich

Lipid Uptake Inhibitor, ML278

Synonym(s):

Lipid Uptake Inhibitor, ML278, 3,5-Dimethoxy-N-(4-(1-(2-methoxyacetyl)indolin-5-yl)thiazol-2-yl)benzamide, SR-BI inhibitor ML278

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$187.00

$187.00


Estimated to ship on16 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$187.00

About This Item

Empirical Formula (Hill Notation):
C23H23N3O5S
CAS Number:
Molecular Weight:
453.51
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

$187.00


Estimated to ship on16 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

6 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, non-toxic, indolinyl-thiazole based compound that acts as a highly potent, reversible inhibitor of lipid uptake via scavenger receptor class B type I (SR-BI; IC50 = 6 nM). Shown to block the uptake of [3H] labeled cholesteryl oleate ester ([3H]CE) from [3H]CE-HDL (IC50 = 7 nM). Exhibits high selectivity when tested against a panel of 67 different receptors and secondary targets (at 10 µM). Displays excellent stability in human plasma (99% remaining after 5 h, with 94% plasma protein bound) and mouse liver microsomes (75% remaining after 1 h).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
scavenger receptor class B type I
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Dockendorff, C., et al. 2015. ACS Med. Chem. Lett.6, 375.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service