Skip to Content
Merck
All Photos(1)

Key Documents

5.38241

Sigma-Aldrich

Nur77-p38 Interaction Inhibitor, PDNPA

Synonym(s):

Nur77-p38 Interaction Inhibitor, PDNPA, Pentyl 2-(3,5-dihydroxy-2-nonanoylphenyl)acetate, NR4A1 Inhibitor, PDNPA, TR3-p38 Interaction Inhibitor, PDNPA

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$360.00

$360.00


Estimated to ship on21 May 2025



Select a Size

Change View
10 MG
$360.00

About This Item

Empirical Formula (Hill Notation):
C22H34O5
Molecular Weight:
378.50
UNSPSC Code:
12352200
NACRES:
NA.77

$360.00


Estimated to ship on21 May 2025


Assay

≥97% (HPLC)

Quality Level

form

solid

potency

1.6 μM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell-permeable phenylacetate based compound that specifically blocks the Nur77-p38α interaction by directly targeting the ligand-binding domain of Nur77 (~1 to 10 µM) and impedes Nur77 phosphorylation by p38α. Does neither modulate p38α kinase activity nor affects the stability of the p38α-Nur77 complex. Inhibits p65 binding to DNA and blocks LPS-induced (100 ng/ml) NF-κB activity in RAW264.7 cells (IC50 = 1.6 µM), but does not affect the interaction between Nur77 and p65. Shown to reduce IL-6 concentration and prolong the lifespan of wild type mice treated with LPS. Also increases the survival rate of wild-type mice subjected to cecal ligation and puncture (CLP)-induced sepsis.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Nur77
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Li, L., et al. 2015. Nat. Chem. Biol.11, 339.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service