Skip to Content
Merck
All Photos(2)

Key Documents

5.38005

Sigma-Aldrich

BHPI

Synonym(s):

BHPI, 3,3- bis(4-Hydroxyphenyl)-7-methyl-1,3-dihydro-2H-indol-2-one, Estrogen Receptor α Inhibitor, 3,3-bis(4-Hydroxyphenyl)-7-methyl-1,3-dihydro-2H-indol-2-one, Estrogen Receptor α Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$158.00

$158.00


Estimated to ship on23 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$158.00

About This Item

Empirical Formula (Hill Notation):
C21H17NO3
CAS Number:
Molecular Weight:
331.36
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$158.00


Estimated to ship on23 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

CC1=C2C(=CC=C1)C(C(=O)N2)(C3=CC=C(C=C3)O)C4=CC=C(C=C4)O

InChI key

FABLAHMQSQFDHR-UHFFFAOYSA-N

General description

A cell-permeable, bioavailable, non-toxic dihydroindolone derived compound that directly interacts with ERα and noncompetitively inhibits E2-ERα regulated gene expression. Selectively arrests the proliferation of therapy resistant ERα+ cancer cells (0.1 - 1.0 µM; IC50 = 15 nM for T47D cells) with no effect on ERα- cells. Shown to hyperactivate PLCγ, deplete endoplasmic reticulum calcium store, elicit massive UPR, and inhibit protein synthesis continually. Downregulates chaperones synthesis and sustains UPR activated cell death. Rapidly induces tumor regression (>60% inhibition) in mouse MCF-7 xenografts (15 mg/kg, i.p., q.d.).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ERα
Reversible: yes

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Andruska, N.D., et al. 2015. Proc. Natl. Acad. Sci. USA.112, 4737.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service