Skip to Content
Merck
All Photos(1)

Key Documents

5.34377

Sigma-Aldrich

CMB-087229

Synonym(s):

CMB-087229, 5-[(3,5-Dichlorophenoxy)methyl]-1,2-dihydro-3H-pyrazol-3-one, CMB087229

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$266.00

$266.00


Estimated to ship on02 June 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
25 MG
$266.00

About This Item

Empirical Formula (Hill Notation):
C10H8Cl2N2O2
CAS Number:
Molecular Weight:
259.09
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$266.00


Estimated to ship on02 June 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

C1=C(C=C(C=C1Cl)Cl)OCC2=CC(=O)NN2

General description

A cell-permeable, blood-brain barrier permeable, non-toxic pyrazolone compound that potently blocks mutant Cu/Zn SOD1 cytotoxicity (EC50 = 67 nM in PC12 cells expressing mutant G93A SOD1). Shown to directly interact with 26S proteasome PSMC1 and PSMC4 and the chaperone protein TCP-1 and enhance proteasome activity. Offers neuroprotection against protein aggregation induced toxicity and prolongs the survival of ALS transgenic mice by 13.3% (20 mg/kg, i.p., q.d.) Demonstrates attractive microsomal (T1/2 = 93 & 36 min in human & mouse, respectively) and metabolic stability, aqueous solubility (250 µM) and PK profile in mice. Poorly affects the activities of 68-enzymes & receptors, hERG and Cyp450 isozymes at 10 µM.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
26S proteasome PSMC1 and PSMC4 and the chaperone protein TCP-1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tripper, P.C., et al. 2014. ACS Chem. Neurosci.5, 823.
Chen, T., et al. 2012. J. Med. Chem.55, 515.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service