Skip to Content
Merck
All Photos(1)

Key Documents

5.34360

Sigma-Aldrich

Autophagy Inhibitor VII

Synonym(s):

Autophagy Inhibitor VII, 6-Chloro-N-(1-ethylpiperidin-4-yl)-1,2,3,4-tetrahydroacridin-9-amine, Acridinamine 33, Tetrahydroacridine 33, THA33

Sign Into View Organizational & Contract Pricing

Select a Size

5343600001
$353.00

$353.00


Estimated to ship on29 May 2025



Select a Size

Change View
5343600001
$353.00

About This Item

Empirical Formula (Hill Notation):
C20H26ClN3
Molecular Weight:
343.89
UNSPSC Code:
12352200
NACRES:
NA.77

$353.00


Estimated to ship on29 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white to pale yellow

solubility

DMSO: 1.6 mg/mL
water: 50 mg/mL

storage temp.

−20°C

General description

A cell-permeable tetrahydroacridinamine (THA) derived compound that potently suppresses autophagy (EC = 250 nM) with reduced toxicity (LD50 = 27 µM) in U2OS cells. Shown to induce lysosome deacidification and increase LC3-II protein accumulation. Displays attractive biophysical properties.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as HCl salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, T., et al. 2015. J. Med. Chem.58, 3025.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service