Skip to Content
Merck
All Photos(1)

Key Documents

5.34350

Sigma-Aldrich

CDC-like Kinase Inhibitor, Cpd-2

Synonym(s):

CDC-like Kinase Inhibitor, Cpd-2, CLK Inhibitor, Cpd-2

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$234.00

$234.00


Estimated to ship on25 May 2025



Select a Size

Change View
5 MG
$234.00

About This Item

Empirical Formula (Hill Notation):
C20H20N6O
Molecular Weight:
360.41
UNSPSC Code:
12352200
NACRES:
NA.77

$234.00


Estimated to ship on25 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light pink

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable imidazo[1,2-b]pyrazol derived compound that acts as a highly potent and specific inhibitor of CDC-like kinases (CLK; IC50 = 1.1 and 2.4 nM for CLK1 and CLK2, respectively). Exhibits only trivial activity against serine-arginine protein kinases (SRPK) 1, 2, and 3 (IC50 = 200, 310, and 230 nM, respectively). Induces pre-mRNA splicing alteration of multiple genes involved in growth and survival pathways resulting in the depletion of respective proteins. Causes a significance decrease in endogenous phosphorylated serine-arginine proteins and enlarges nuclear speckles in MDA-MB-468 cells. Induces apoptosis and Inhibits the growth of multiple cancer cell lines (GI50 = 3.0, 1.9, 1.7, 2.2, 1.4, 2.0, and 0.6 µM for MDA-MB-468, A549, COLO205, HCT116, NCIH23, SW620, and COLO320 DM cells, respectively).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
CLK1
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Araki, S., et al. 2015. Plos One.10, e0116929.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service