Skip to Content
Merck
All Photos(1)

Key Documents

5.34349

Sigma-Aldrich

ABHD16A Inhibitor, KC01

Synonym(s):

ABHD16A Inhibitor, KC01, (Z)-6-(2-oxo-4-tridecyloxetan-3-ylidene)hexanamide, BAT5 Inhibitor, KC01

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$338.00

$338.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
$338.00

About This Item

Empirical Formula (Hill Notation):
C22H39NO3
Molecular Weight:
365.55
UNSPSC Code:
51111800
NACRES:
NA.77

$338.00


Estimated to ship on29 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

storage temp.

−20°C

General description

A cell-permeable beta-lactone based compound that acts as a potent, selective, and covalent inhibitor of phosphatidylserine (PS) lipase ABHD16A (BAT5, IC50 = 90 and 520 nM for human and mouse ABHD16A, respectively). Displays reduced effect on ~60 other serine hydrolases profiled. Shown to block the PS lipase activity of membrane fractions from COLO205, K562 and MCF7 cell lines and is also shown to diminish the PS lipase activity of brain membrane lysates from Abhd12+/+ (IC50 = 290 nM) and Abhd12-/- (IC50 = 340 nM) mice. Significantly reduces lysophosphatidylserine (lyso-PSs) levels in cells, without affecting the levels of other lipids, such as phosphatidylcholine (PC), phosphatidylethano-lamines (PE), lyso-PCs, lyso-PEs or monoacylglycerols. Blunts lipopolysaccharide-induce release of IL-6, IL-1β in mouse macrophages.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ABHD16A

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Kamat, S., et al. 2015. Nat. Chem. Biol.11, 164.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service