Skip to Content
Merck
All Photos(1)

Key Documents

5.34337

Sigma-Aldrich

CADA

≥98% (HPLC), solid, HIV inhibitor, Calbiochem®

Synonym(s):

HIV Inhibitor, CADA, 1-Benzyl-7-methylene-5,9-ditosyl-5,9-diaza-azoniacyclododecan-1-ium chloride, CD4 Inhibitor, CADA

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$382.00

$382.00


In StockDetails



Select a Size

Change View
10 MG
$382.00

About This Item

Empirical Formula (Hill Notation):
C31H39N3O4S2 · xHCl
Molecular Weight:
581.79 (free base basis)
UNSPSC Code:
51111800
NACRES:
NA.77

$382.00


In StockDetails


Product Name

HIV Inhibitor, CADA,

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
water: insoluble

storage temp.

−20°C

General description

A cell-permeable macrocyle cyclotriazadisulfonamide derivative compound that inhibits HIV replication (IC50 = 300 nM to 3.3 µM) and entry into cells by blocking co-translational translocation of nascent CD4 across endoplasmic reticulum membranes. Its action appears to be highly selective in reducing the expression of human CD4 and it does not affect the level of any other cell surface marker studied. Does not affect CD4 mRNA levels in cells. Shown to be effective against HIV-1 subtypes A to H, AE, and O and human herpesvirus 7 (HHV-7; IC50, 300 nM to 1.5 µM) in T-cell lines and PBMCs. Also reported to block X4 HIV-1 NL4.3 and R5 SIVmac251 laboratory strains infection in human T cells. Can act synergistically with cellulose acetate phthalate (CAP) and other anti-HIV drugs to inhibit HIV-1 and SIV infections.

Biochem/physiol Actions

Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Vermeire, K., et al. 2008. Curr. HIV Res.6, 246.
Bell, T. W., et al. 2006. J. Med. Chem.49, 1291.
Vermeire, K., et al. 2004. AIDS.18, 2115.
Vermeire, K., et al. 2002. Virology.302, 342.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service