Skip to Content
Merck
All Photos(1)

Key Documents

5.33966

Sigma-Aldrich

LDH-A Inhibitor III, NHI-1

Synonym(s):

LDH-A Inhibitor III, NHI-1, 1-Hydroxy-6-phenyl-4-trifluoromethyl-1H-indole-2-carboxylic acid, Lactate Dehydrogenase A Inhibitor III, LDH-M Inhibitor III

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$363.00

$363.00


Estimated to ship on30 May 2025



Select a Size

Change View
10 MG
$363.00

About This Item

Empirical Formula (Hill Notation):
C16H10F3NO3
CAS Number:
Molecular Weight:
321.25
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$363.00


Estimated to ship on30 May 2025


Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, non-cytotoxic N-hydroxyindole (NHI) carboxylate that acts as a potent, reversible, and isoform selective inhibitor of lactic dehydrogenase (LDH) A/LDH-1. Inhibition appears to be competitive with respect to both pyruvate (Ki = 4.7 µM) and NADH (cofactor; Ki = 8.9 µM). Acts by occupying the whole substrate pocket and part of the cofactor pocket of LDH-A. Blocks the growth of various cancer cell lines by causing an arrest at G1 phase and inducing apoptosis (IC50 = 10.8, 10.6, 11.4, and 31.5 µM for A2780/cOHP, MSTO-211H, NIH-H28, and H630 cells, respectively). Exhibits enhanced anticancer activity under hypoxic conditions (GI50 = 0.90 µM/hypoxia vs. 16.3 µM/normoxia in LPC006 cultures) and strongly synergizes with gemcitabine (Cat. No. 504594).Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
LDH-A/LDH-1
Target IC50: 10.8, 10.6, 11.4, and 31.5 &micro

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Maftouh, M., et al. 2014. Br. J. Cancer.110, 172.
Granchi, C., et al. 2011. J. Med. Chem.54, 1599.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service