5.33796
Asparagine Endopeptidase Inhibitor, AENK
Synonym(s):
Asparagine Endopeptidase Inhibitor, AENK, Fmoc-Ala-Glu-Asn-Lys-NH2, Asparagine Endopeptidase Inhibitor, AENK, AEP Inhibitor, AENK
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Assay
≥98% (HPLC)
Quality Level
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
white
solubility
water: 25 mg/mL
storage temp.
−20°C
General description
An amino and carboxy-terminal blocked tetra peptide that acts as a specific, competitive inhibitor of the protease activity of asparagine endopeptidase (AEP), an enzyme that cleaves substrates on the C-terminal side of asparagine. Believed to act as a competitive substrate that itself is hydrolyzed by AEP. Shown to block processing of tetanus toxin antigen (TTCF) in vitro (~1 mg/ml) and retard TTCF presentation to T cells in vivo. Does not affect the cleavage of cathepsin B/L and cathepsin S substrates. Shown to selectively and completely suppress tau cleavage by AEP (~ 1 mg/ml).
An amino and carboxy-terminal blocked tetra peptide that acts as a specific, competitive inhibitor of the protease activity of asparagine endopeptidase (AEP), an enzyme that cleaves substrates on the C-terminal side of asparagine. Believed to act as a competitive substrate that itself is hydrolyzed by AEP. Shown to block processing of tetanus toxin antigen (TTCF) in vitro (~1 mg/ml) and retard TTCF presentation to T cells in vivo. Does not affect the cleavage of cathepsin B/L and cathepsin S substrates. Shown to selectively and completely suppress tau cleavage by AEP (~ 1 mg/ml).
Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Biochem/physiol Actions
Primary Target
AEP
AEP
Reversible: yes
Packaging
Packaged under inert gas
Warning
Toxicity: Standard Handling (A)
Sequence
Fmoc-Ala-Glu-Asn-Lys-NH2
Physical form
Supplied as a TFA salt.
Reconstitution
Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Other Notes
Zhang, Z., et al. 1999. Nat. Med.20, 1254.
Monoury, B., et al. 1998. Nat.396, 695.
Monoury, B., et al. 1998. Nat.396, 695.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service