Skip to Content
Merck
All Photos(1)

Key Documents

5.33063

Sigma-Aldrich

VPS34 Inhibitor, SAR405

Synonym(s):

VPS34 Inhibitor, SAR405, Vacuolar Protein Sorting 34 Inhibitor, SAR405, (S)-1-((5-chloropyridin-3-yl)methyl)-8-((R)-3-methylmorpholino)-2-(trifluoromethyl)-3,4-dihydro-1H-pyrimido[1,2-a]pyrimidin-6(2H)-one

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$296.00

$296.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$296.00

About This Item

Empirical Formula (Hill Notation):
C19H21ClF3N5O
CAS Number:
Molecular Weight:
427.85
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$296.00


Estimated to ship on24 May 2025


Request a Bulk Order

Assay

≥96% (HPLC)

Quality Level

form

powder

potency

1.5 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

−20°C

SMILES string

FC(F)(F)[C@H]1N(c3[n]([c](cc(n3)N4[C@@H](COCC4)C)=O)CC1)Cc2cncc(c2)Cl

InChI

1S/C19H21ClF3N5O2/c1-12-11-30-5-4-26(12)16-7-17(29)27-3-2-15(19(21,22)23)28(18(27)25-16)10-13-6-14(20)9-24-8-13/h6-9,12,15H,2-5,10-11H2,1H3/t12-,15+/m1/s1

InChI key

SPDQRCUBFSRAFI-DOMZBBRYSA-N

General description

A cell-permeable pyrimido[1,2-a]pyrimidin-6(2H)-one derivative that acts as a potent, selective, and ATP competitive inhibitor of vacuolar protein sorting 34 kinase activity (Vps34; Kd = 1.5 nM, IC50 = 1.2 nM in a biochemical assay, and IC50 = 27 nM in a cell based assay). Does not affect the activity of over 200 other protein kinases including class I and II PI 3 kinases and mTOR (~ 10 µM). However, it displays a weak inhibitory activity against SMG1 at higher concentrations (IC50 = 1 µM). Does not affect the Akt phosphorylation even at higher concentrations (~10 µM), Shown to affect the late endosome-lysosome compartment formation and block the formation of autophagosomes in starved HeLa and H1299 cells (~ 1 µM). Acts synergistically with everolimus to block the proliferation of ACHN and 786-O renal tumor cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
VPS34

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ronan, B., et al. 2014. Nat. Chem. Biol.10, 1013.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service