Skip to Content
Merck
All Photos(1)

Documents

5.32910

Sigma-Aldrich

TrxR Inhibitor, D9

Synonym(s):

TrxR Inhibitor, D9, Thioredoxin inhibitor, D9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20AuOPS
Molecular Weight:
596.43
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

−20°C

General description

A cell permeable, highly stable phosphinyl gold (I) acetylide complex that acts as a highly potent inhibitor of thioredoxin reductase (TrxR; EC50 = 2.8 nM). Acts by blocking the redox-active selenenylsulfide motif of TrxR in an irreversible manner. Displays high selectivity over glutathione reductase (EC50 >1000 nM). Selectively reduces the viability and proliferation of MCF-7 and HT-29 cancer cells (IC50 = 30 and 100 nM, respectively) without affecting normal cells. Suppresses the growth of MCF-7 xenografts in BALB/c nude mice (5 mg/kg; i.v.,given on alternate days for 15 days).
A cell permeable, highly stable phosphinyl gold (I) acetylide complex that acts as a highly potent inhibitor of thioredoxin reductase (TrxR; EC50 = 2.8 nM). Acts by blocking the redox-active selenenylsulfide motif of TrxR in an irreversible manner. Displays high selectivity over glutathione reductase (EC50 >1000 nM). Selectively reduces the viability and proliferation of MCF-7 and HT-29 cancer cells (IC50 = 30 and 100 nM, respectively) without affecting normal cells. Suppresses the growth of MCF-7 xenografts in BALB/c nude mice (5 mg/kg; i.v.,given on alternate days for 15 days).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
TrxR Inhibitor, D9, is a cell permeable, highly stable phosphinyl gold (I) acetylide complex that acts as a highly potent inhibitor of thioredoxin reductase (TrxR; EC₅₀ = 2.8 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
TrxR
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, D., et al. 2014. J. Med. Chem.57, 8132.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service