Skip to Content
Merck
All Photos(1)

Key Documents

5.32722

Sigma-Aldrich

Nurr1 Activator, IP7e

Synonym(s):

Nurr1 Activator, IP7e, (6-(4-((2-Methoxyethoxy)methyl)phenyl)-5-methyl-3-phenylisoxazolo(4,5-c)pyridin-4(5H)-one), 6-(4-((2-Methoxyethoxy)methyl)phenyl)-5-methyl-3-phenyl[1,2]oxazolo[4,5-c]pyridin-4(5H)-one, Isoxazolo-Pyridinone 7e, NR4A2 Activator

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$232.00

$232.00


Estimated to ship on18 April 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$232.00

About This Item

Empirical Formula (Hill Notation):
C23H22N2O4
CAS Number:
Molecular Weight:
390.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$232.00


Estimated to ship on18 April 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable, blood-brain barrier permeable, potent activator of Nurr1/NR4A2-dependent transcription activity in (EC₅₀ = 3.9 nM in MN90 cell line).
A cell-permeable, isoxazolopyridinone compound that acts as a potent activator of Nurr1/NR4A2-dependent transcription activity in reporter assays using Nurr1-expressing murine midbrain dopaminergic neuronal cell line MN9D (EC50 = 3.9 nM, Emax = 2.1-fold of basal activity) and exhibits oral availability as well as blood-brain-barrier permeability in mice. Preventive treatment (10 mg/kg/12h from 7th to 23rd day post MOG35-55 immunization) is shown to delay the onset (15 d.p.i. with & 12 d.p.i. without preventive treatment) and severity of experimental autoimmune encephalomyelitis (EAE) in a murine multiple sclerosis (MS) model in vivo, while no improvement of EAE symptoms is observed if the treatment starts after the disease onset (10 mg/kg/12h from 21 d.p.i. to 36 d.p.i.). Consistent with the known negative regulating role of Nurr1 against NF-κB pathway, preventive treatment is shown to result in significantly reduced expression of 16 NF-κB pathway genes in the spinal cord of EAE mice.
A cell-permeable, isoxazolopyridinone compound that acts as a potent activator of Nurr1/NR4A2-dependent transcription activity in reporter assays using Nurr1-expressing murine midbrain dopaminergic neuronal cell line MN9D (EC50 = 3.9 nM, Emax = 2.1-fold of basal activity) and exhibits oral availability as well as blood-brain-barrier permeability in mice. Preventive treatment (10 mg/kg/12h from 7th to 23rd day post MOG35-55 immunization) is shown to delay the onset (15 d.p.i. with & 12 d.p.i. without preventive treatment) and severity of experimental autoimmune encephalomyelitis (EAE) in a murine multiple sclerosis (MS) model in vivo, while no improvement of EAE symptoms is observed if the treatment starts after the disease onset (10 mg/kg/12h from 21 d.p.i. to 36 d.p.i.). Consistent with the known negative regulating role of Nurr1 against NF-κB pathway, preventive treatment is shown to result in significantly reduced expression of 16 NF-κB pathway genes in the spinal cord of EAE mice.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Nurr1/NR4A2

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Montarolo, F., 2014. PLoS One.9, e108791.
Hintermann, S., et al. 2007. Bioorg. Med. Chem. Lett.17, 193.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service