Skip to Content
Merck
All Photos(1)

Key Documents

5.32586

Sigma-Aldrich

AEBSF hydrochloride

≥99% (HPLC), powder, serine protease inhibitor, Calbiochem®

Synonym(s):

AEBSF, HCl ≥99% by HPLC, 4-(2-Aminoethyl)benzenesulfonylfluoride, HCl, 4-(2-Aminoethyl)benzene-1-sulfonyl fluoride, HCl

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$297.00

$297.00


Estimated to ship on18 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
250 MG
$297.00

About This Item

Empirical Formula (Hill Notation):
C8H10FNO2S · xHCl
CAS Number:
Molecular Weight:
203.23 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$297.00


Estimated to ship on18 May 2025

New, lower price on this item!

Request a Bulk Order

Product Name

AEBSF, HCl ≥99% by HPLC,

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

H2O: 50 mg/mL

storage temp.

2-8°C

SMILES string

F[S](=O)(=O)c1ccc(cc1)CCN.[Cl-].[H+]

InChI

1S/C8H10FNO2S.ClH/c9-13(11,12)8-3-1-7(2-4-8)5-6-10;/h1-4H,5-6,10H2;1H

InChI key

WRDABNWSWOHGMS-UHFFFAOYSA-N

General description

AEBSF >99%; 30827-99-7
Specific irreversible inhibitor of serine proteases. Reacts covalently with a serine at the active site. Inhibits chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. Stable, nontoxic alternative to PMSF (Cat. No. 52332). Used at the same molar concentration (0.1-1.0 mM) as PMSF for most applications. Exhibits low inhibitory activity towards proteinase K. A material with ≥97% purity by HPLC (Cat. No. 101500) & its InSolution is also available (Cat. No. 508436).Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Primary Target
Serine proteases
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Corrosive (L)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Citron, M., et al. 1996. Neuron17, 171.

Ohkubo, I., et al. 1994. J. Biochem.116, 1182.

Mintz, G.R. 1993. BioPharm.6, 34.

Lawson, W.B., et al. 1982. Folia Haematol.109, 52.

Markwardt, F., et al. 1974. Biochem. Pharmacol.23, 2247.
Markwardt, F., et al. 1973. Thromb. Res.2, 343.
Baker, B.R., and Cory, M. 1971. J. Med. Chem.14, 119.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service