Skip to Content
Merck
All Photos(1)

Key Documents

5.31889

Sigma-Aldrich

STING Agonist, 2′3′-cGAMP

≥98% (HPLC), solid, STING agonist, Calbiochem®

Synonym(s):

STING Agonist, 2′3′-cGAMP, c[G(2ʹ,5ʹ)pA(3ʹ,5ʹ)p], 2ʹ,5ʹ-3ʹ,5ʹ-cGAMP

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
$513.00

$513.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
500 μG
$513.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$513.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Product Name

STING Agonist, 2′3′-cGAMP,

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

4 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

H2O: soluble

storage temp.

−20°C

SMILES string

[P]1(=O)(O[C@@H]2[C@H](O[C@H]([C@@H]2O)[n]6c7ncnc(c7nc6)N)CO[P](=O)(O[C@H]3[C@@H](O[C@@H]([C@H]3O)CO1)[n]4c5[nH]c(n[c](c5nc4)=O)N)O)O

InChI

1S/C20H24N10O13P2/c21-14-8-15(24-3-23-14)29(4-25-8)18-11(32)12-7(41-18)2-39-45(36,37)43-13-10(31)6(1-38-44(34,35)42-12)40-19(13)30-5-26-9-16(30)27-20(22)28-17(9)33/h3-7,10-13,18-19,31-32H,1-2H2,(H,34,35)(H,36,37)(H2,21,23,24)(H3,22,27,28,33)/t6-,7-,10-,11-,12-,13-,18-,19-/m1/s1

InChI key

XRILCFTWUCUKJR-INFSMZHSSA-N

General description

A cyclic-dinucleotide (cyclic-GMP-AMP) based compound that binds to the C-terminal domain of adaptor endoplasmic reticulum protein known as STING with high affinity (Kd = 4 nM) and induces the synthesis of interferon-b (IFN-b) in mammalian cells (EC50 = 20 nM). Shown to increase IFN-b production in M2-polarized macrophages associated with non-small cell lung carcinoma and enhances the expression of M1 markers iNOS and IL-12p40, while simultaneously reducing the expression of the M2 markers, Arg-1 and Fizz1. Exhibits about 300 fold greater affinity for STING than c-DiGMP, 3′,2′-cGAMP, and 3′,3′-cGAMP and about 75 fold greater affinity over 2′,2′-cGAMP.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
STING

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wu, X., et al. 2014. Nuc. Aci. Res.42, 8243.
Zhang, X., et al. 2013. Mol. Cell.51, 226.
Wu, J., et al. 2013. Science.339, 26.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service