Skip to Content
Merck
All Photos(1)

Key Documents

5.31369

Sigma-Aldrich

RORγt Inverse Agonist II, GSK805

Synonym(s):

RORγt Inverse Agonist II, GSK805, N-(3,5-Dichloro-4-(2-(trifluoromethoxy)phenyl)phenyl)-2-(4-ethylsulfonylphenyl)acetamide, N-(2,6-Dichloro-2ʹ-(trifluoromethoxy)biphenyl-4-yl)-2-(4-(ethylsulfonyl)phenyl)acetamide, Retinoid-related Orphan Receptor γ t Antagonist II, RORγt A

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H18Cl2F3NO4S
CAS Number:
Molecular Weight:
532.36
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)Oc1c(cccc1)c2c(cc(cc2Cl)NC(=O)Cc3ccc(cc3)[S](=O)(=O)CC)Cl

General description

A cell-permeable, orally available, non-toxic, conformationally restricted biaryl-amide compound that acts as a potent retinoid-related orphan receptor γt (RORγt) isoform selective inverse agonist and suppresses T helper 17 (Th17) responses, including IL-17 and IFN-γ production (~ 500 nM). Directly and reversibly interacts with the ligand binding domain of RORγt to block its transcriptional effects, however, it does not affect RORγt binding to DNA. Induces RORγt binding to Gata3 resulting in increased expression of GATA3. Reduces the severity of autoimmune encephalomyelitis in murine models of multiple sclerosis (30 mg/kg/day, p.o.).
A cell-permeable, orally available, non-toxic, conformationally restricted biaryl-amide compound that acts as a potent retinoid-related orphan receptor γt (RORγt) isoform selective inverse agonist and suppresses T helper 17 (Th17) responses, including IL-17 and IFN-γ production (~ 500 nM). Directly and reversibly interacts with the ligand binding domain of RORγt to block its transcriptional effects, however, it does not affect RORγt binding to DNA. Induces RORγt binding to Gata3 resulting in increased expression of GATA3. Reduces the severity of autoimmune encephalomyelitis in murine models of multiple sclerosis (30 mg/kg/day, p.o.).

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
RORγt
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Xiao, S., et al. 2014. Immunity.40, 477.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service