Skip to Content
Merck
All Photos(1)

Key Documents

5.31091

Sigma-Aldrich

Porcn Inhibitor IV, LGK974

Synonym(s):

Porcn Inhibitor IV, LGK974, 2-(2ʹ,3-Dimethyl-2,4ʹ-bipyridin-5-yl)-N-(5-(pyrazin-2-yl)pyridin-2-yl)acetamide, Wnt Pathway Inhibitor XXIII, LGK-974

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$295.00

$295.00


Estimated to ship on22 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$295.00

About This Item

Empirical Formula (Hill Notation):
C23H20N6O
CAS Number:
Molecular Weight:
396.44
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$295.00


Estimated to ship on22 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

N(c3ncc(cc3)c4nccnc4)C(=O)Cc1cnc(c(c1)C)c2cc(ncc2)C

InChI

1S/C23H20N6O/c1-15-9-17(12-28-23(15)18-5-6-25-16(2)10-18)11-22(30)29-21-4-3-19(13-27-21)20-14-24-7-8-26-20/h3-10,12-14H,11H2,1-2H3,(H,27,29,30)

InChI key

XXYGTCZJJLTAGH-UHFFFAOYSA-N

General description

A cell-permeable C59 (Cat. No. 500496) structural analog that potently inhibits Wnt secretion from L-cell Wnt1/2/3/3a/6/7a/9a transfectants (IC50 from 50 pM to 1.1 nM) via direct porcupine (Porcn), but not Wntless, affinity interaction. Shown to inhibit the proliferation of head and neck squamous cell carcinoma (HNSCC) line HN30 (IC50 = 0.3 nM), but not 293T even at a high concentration of 20 µM. In mice & rats receiving therapeutic LGK974 oral dosages (up to 5 mg/10 mL/kg/d), a delayed tissue Wnt signaling suppression (max inhibition 7 to 10 h post dosing) is seen due to pre-existing Wnt in tissue, followed by a full recovery of tissue Wnt signalling at 24 h upon drug clearance. The periodic daily inhibition/recovering cycle is shown to result in tumor regression in mice & rats, while allowing Wnt signaling-dependent normal tissue homeostasis. Overdosing is reported to result in loss of intestinal epithelium in rats receiving 14 consecutive 20 mg/kg daily oral dosages.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Porcupine
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Liu, J., et al. 2013. Proc. Natl. Acad. Sci. USA110, 20224.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service