Skip to Content
Merck
All Photos(1)

Key Documents

5.31068

Sigma-Aldrich

Lenaldekar (LDK)

Synonym(s):

Lenaldekar (LDK), T Cell Proliferation Inhibitor, LDK

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$207.00

$207.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$207.00

About This Item

Empirical Formula (Hill Notation):
C18H14N4
CAS Number:
Molecular Weight:
286.33
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

$207.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[nH]1c2c(c(c1)\C=N\Nc3c4ncccc4ccc3)cccc2

InChI

1S/C18H14N4/c1-2-8-16-15(7-1)14(11-20-16)12-21-22-17-9-3-5-13-6-4-10-19-18(13)17/h1-12,20,22H/b21-12+

InChI key

QLMFOQYHZJSYJF-CIAFOILYSA-N

General description

A quinolinyl hydrazone derived compound that specifically blocks the proliferation of T cells in a dose-dependent manner (IC50 = 3 µM in CD3+, CD4+, and CD8+ cells) without affecting other cell types. Its anti-proliferative effects are non-cytolytic in nature. Induces cell cycle delay at G2/M phase. Selectively induces apoptosis in human leukemic blasts and is shown to be active against all acute lymphoblastic leukemia cell lines (IC50 = 1 -2 µM) and also displays in vivo activity in a zebrafish T-ALL model. Reduces the growth of human T-ALL in murine xenograft models (16 mg/kg, i,p, b.i.d). Also shown to specifically block myelin specific T cell responses and reduce inflammation and demyelination in experimental autoimmune encephalomyelitis, possibly via its interactions with insulin-like growth factor-1 receptor. Diminishes the phosphorylation of T308 and S473 on Akt, however, its action is believed to be upstream of PI-3K/Akt. Does not affect the activity of over 450 other kinases in any significant manner. Exhibits a plasma t1/2 = 2 h.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes
Target IC50: 3 µM to block proliferation of T lymphocytes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Cusick, M.F., et al. 1999. PNAS.96, 12192.
Ridges, S, et al. 2012. Blood.119, 5621.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service