Skip to Content
Merck
All Photos(1)

Key Documents

5.31042

Sigma-Aldrich

ICI 182,780

≥98% (HPLC), powder, GPR30 agonist, Calbiochem®

Synonym(s):

Estrogen Receptor Antagonist, ICI 182,780, GPR30 Agonist, ICI 182,780, ICI 182780, ICI182780

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$305.00

$305.00


Estimated to ship on22 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$305.00

About This Item

Empirical Formula (Hill Notation):
C32H47F5O3S
CAS Number:
Molecular Weight:
606.77
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$305.00


Estimated to ship on22 May 2025

New, lower price on this item!

Request a Bulk Order

Product Name

Estrogen Receptor Antagonist, ICI 182,780,

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)C(F)(F)CCC[S](=O)CCCCCCCCC[C@H]1[C@H]2[C@H]3[C@@]([C@H](CC3)O)(CC[C@@H]2c4c(cc(cc4)O)C1)C

InChI

1S/C32H47F5O3S/c1-30-17-15-26-25-12-11-24(38)21-23(25)20-22(29(26)27(30)13-14-28(30)39)10-7-5-3-2-4-6-8-18-41(40)19-9-16-31(33,34)32(35,36)37/h11-12,21-22,26-29,38-39H,2-10,13-20H2,1H3/t22-,26-,27+,28+,29-,30+,41?/m1/s1

InChI key

VWUXBMIQPBEWFH-WCCTWKNTSA-N

General description

A cell-permeable estradiol derivative that acts as an estrogen antagonist and down-regulates estrogen receptors without affecting estrogen receptor-mediated progesterone receptor expression. Hence, it lacks estrogen agonist activity. Shown to be more effective than tamoxifen in reducing estrogen receptors in breast tumor cells. Suppresses the growth of MCF-7 xenografts in mice for twice as long as tamoxifen and delays tumor growth.



Please note that the molecular weight for this compound is batch-specific due to variable water content.
A cell-permeable estrogen antagonist. More effective than tamoxifen in reducing estrogen receptors in breast tumor cells.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
ER
Secondary Target
GPR30

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wakeling, A. E., et al. 1991. Cancer Res.51, 3867.
Dauvois, S., et al. 1993. J. Cell. Sci.106, 1377.
Howell, A., et al. 1995. The Lancet.8941, 29.
Robertson, J.F., et al. 2001. J. Steroid Biochem. Mol Biol.79, 209.

Legal Information

Sold with the permission of AstraZeneca UK. Limited for research purposes only.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service