Skip to Content
Merck
All Photos(1)

Key Documents

5.30154

Sigma-Aldrich

BMI-1 Expression Inhibitor, PTC-209

Synonym(s):

BMI-1 Expression Inhibitor, PTC-209, FLVI2/BMI1 Expression Inhibitor, MGC12685 Expression Inhibitor, PCGF4 Expression Inhibitor, PTC209; RNF51 Expression Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$279.00

$279.00


Estimated to ship on31 May 2025

New, lower price on this item!


Select a Size

Change View
10 MG
$279.00

About This Item

Empirical Formula (Hill Notation):
C17H13Br2N5OS
CAS Number:
Molecular Weight:
495.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$279.00


Estimated to ship on31 May 2025

New, lower price on this item!

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

[s]1c(nc(c1)c3[n]4c(nc3C)nccc4)Nc2c(cc(cc2Br)OC)Br

InChI

1S/C17H13Br2N5OS/c1-9-15(24-5-3-4-20-16(24)21-9)13-8-26-17(22-13)23-14-11(18)6-10(25-2)7-12(14)19/h3-8H,1-2H3,(H,22,23)

InChI key

XVOOCQSWCCRVDY-UHFFFAOYSA-N

General description

A cell-permeable imidazopyrimidinyl-thiazolamine compound that effectively downregulates cellular BMI-1 protein level both in cultures in vitro (IC50 = 0.5 µM/48 h/HT1080 and <0.37 µM/HCT116/overnight) and in mice in vivo (4.5 ng vs.13.7 ng per mg HT1080 tumor tissue, respectively, with or without 10-d 60 mg/kg/d treatment via s.c.) by inhibiting BMI-1 transcription (IC50 = 0.5 µM against overnight reporter transcription in HEK293) via a yet unidentified mechanism, thereby reducing BMI-1-dependent RING1A E3 ligase activity and RING1A-mediated H2A ubiquitination (uH2A)/epigenetic regulations. Shown to exhibit antiproliferation activity against human colon cancer cells both in cultures (IC50<300 nM against LS174T and primary colon cancer cultures) in vitro and in mice (30 to 60 mg/kg/d s.c.) in vivo by inducing cell cycle arrest at G0 and apoptosis. Reduced frequency of sphere-forming cells is observed among surviving cells (by 1.7- to 2.2-fold) upon PTC-209 removal post a 4-day 0.1 µM drug treatment period in primary colon cancer cultures, indicating a greater impact of drug treatment on self-renewing cancer-initiating cells (CICs; cancer stem cells). Likewise, surviving colon cancer cells from tumor-bearing mice at the end of drug treatment are shown to contain reduced population of tumor-initiating cells when xenografted into secondary recipient mice.

Biochem/physiol Actions

Cell permeable: yes
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kreso, A., et al. 2014. Nat. Med.20, 29.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service