528151
PARP Inhibitor VIII, PJ34
InSolution, ≥98%
Synonym(s):
InSolution PARP Inhibitor VIII, PJ34, 2-(Dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide, HCl, N-(6-Oxo-5,6-dihydrophenanthridin-2-yl)-N,N-dimethylacetamide, HCl, N-(6-Oxo-5,6-dihydrophenanthridin-2-yl)-N,N-dimethylacetamide, HCl, 2-(Dimethylamino)-N-(6-oxo-5,6-dihydrophenanthridin-2-yl)acetamide, HCl
Select a Size
$218.00
Estimated to ship on20 April 2025
Select a Size
About This Item
$218.00
Estimated to ship on20 April 2025
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
protect from light
shipped in
ambient
storage temp.
−70°C
SMILES string
O=C(CN(C)C)NC(C=C1)=CC(C2=C3C=CC=C2)=C1NC3=O
General description
Biochem/physiol Actions
PARP
Packaging
Warning
Physical form
Reconstitution
Other Notes
Garcia-Soriano, F.G., et al. 2001. Nature Med.7, 108.
Mabley, G.J., et al. 2001. Inflamm. Res.50, 561.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service