Skip to Content
Merck
All Photos(1)

Key Documents

528108

Sigma-Aldrich

PI 3-Kγ Inhibitor II

The PI 3-Kγ Inhibitor II, also referenced under CAS 648449-76-7, controls the biological activity of PI 3-Kγ. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PI 3-Kγ Inhibitor II, 5-(2,2-Difluoro-benzo[1,3]dioxol-5-ylmethylene)-thiazolidine-2,4-dione, PI 3-K Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$402.00

$402.00


Estimated to ship on22 May 2025



Select a Size

Change View
5 MG
$402.00

About This Item

Empirical Formula (Hill Notation):
C11H5F2NO4S
CAS Number:
Molecular Weight:
285.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$402.00


Estimated to ship on22 May 2025


Quality Level

Assay

≥98% (HPLC)

form

solid

potency

180 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

FC1(Oc2c(ccc(c2)\C=C3\SC(=O)NC\3=O)O1)F

InChI

1S/C11H5F2NO4S/c12-11(13)17-6-2-1-5(3-7(6)18-11)4-8-9(15)14-10(16)19-8/h1-4H,(H,14,15,16)/b8-4+

InChI key

SRLVNYDXMUGOFI-XBXARRHUSA-N

General description

A cell-permeable thiazolidinedione compound that acts as a potent and ATP-competitive inhibitor of PI 3-Kγ (Ki = 180 nM; IC50 = 250 nM). Exhibits great selectivity over PI 3-Kα (IC50 = 4.5 µM), PI 3-Kβ and δ (IC50 >20 µM), and shows little effect towards a large panel of receptors, unrelated enzymes, ion channels, and 38 commonly studied kinases. Shown to block MCP-1-mediated monocyte chemotaxis in vitro and exhibit better in vivo efficacy than LY294002 (Cat. No. 440202 and 440204) in blocking RANTES- or thioglycollate-induced peritoneal neutrophil recruitment by oral administration in a murine peritonitis model.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PI 3-Kγ
Product competes with ATP.
Reversible: no
Target IC50: 250 nM against PI 3-Kγ

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bilancio, A., et al. 2006. Blood107, 642.
Camps, M., et al. 2005. Nat. Med.11, 936.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manoj K Rathinaswamy et al.
eLife, 10 (2021-03-05)
Class I Phosphoinositide 3-kinases (PI3Ks) are master regulators of cellular functions, with the class IB PI3K catalytic subunit (p110γ) playing key roles in immune signalling. p110γ is a key factor in inflammatory diseases and has been identified as a therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service