Skip to Content
Merck
All Photos(1)

Documents

526526

Sigma-Aldrich

PIM3 Kinase Inhibitor VII, M-110

Synonym(s):

PIM3 Kinase Inhibitor VII, M-110, Nʹ-(1-(4-Chloro-2-hydroxyphenyl)propylidene)-2-((3-morpholinopropyl)amino)isonicotinohydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H28ClN5O3
Molecular Weight:
445.94
UNSPSC Code:
12352200
NACRES:
NA.28

Assay

≥97% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable hydroxyphenyl-propylidene-benzohydrazide compound that acts as a potent, ATP-competitive (Ki = 0.3 µM), and highly isoform-selective PIM inhibitor (IC50 = 2.5, 2.5, and 0.047 µM against PIM1, PIM2, and PIM3, respectively, with PIMtide as substrate, [ATP] = 10 µM), while affecting CK2α2 only at much higher concentrations (IC50 = 5 µM, [ATP] = 10 µM) and exhibiting little or no activity against a panel of 258 other kinases (<40% inhibition; [M-110] = 5 µM). M-100 treatment (10 µM for 18 hr) is shown to inhibit both basal (by 73% and 83% of DMSO control in DU-145 prostate cancer and MiaPaCa2 pancreatic cancer cells, respectively) and IL-6-stimulated (by 69% of DMSO control in DU-145 cells) STAT3 phosphorylation on Tyr705, while displaying little effect toward Tyr694 phosphoylation (in 22RV1 cultures). Isoform-specific knockdowns in DU-145 cells likewise identify PIM-3, but not PIM-1 or -2, as the kinase responsible for cellular STAT3 Tyr705 phosphorylation.
A cell-permeable hydroxyphenyl-propylidene-benzohydrazide compound that acts as a potent, ATP-competitive (Ki = 0.3 µM), and highly isoform-selective PIM inhibitor (IC50 = 2.5, 2.5, and 0.047 µM against PIM1, PIM2, and PIM3, respectively; [ATP] = 10 µM), while affecting CK2α2 only at much higher concentrations (IC50 = 5 µM, [ATP] = 10 µM) and exhibiting little or no activity against a panel of 258 other kinases (<40% inhibition at 5 µM). Shown to inhibit PIM3-dependent STAT3 Tyr705 phosphorylation in DU-145 prostate cancer and MiaPaCa2 pancreatic cancer cells (by 73% and 83%, respectively; 10 µM for 18 h), while displaying little effect toward STAT3 Tyr694 phosphoylation in 22RV1 cultures.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C. Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Chang, M., et al. 2010. Mol. Cancer Ther.9, 2478.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mirco Glitscher et al.
Antiviral research, 226, 105891-105891 (2024-04-23)
Zoonoses such as ZIKV and SARS-CoV-2 pose a severe risk to global health. There is urgent need for broad antiviral strategies based on host-targets filling gaps between pathogen emergence and availability of therapeutic or preventive strategies. Significant reduction of pathogen

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service