Skip to Content
Merck
All Photos(1)

Key Documents

526520

Sigma-Aldrich

PIM1 Inhibitor II

Synonym(s):

PIM1 Inhibitor II, 3-Cyano-4-phenyl-6-(3-bromo-6-hydroxy)phenyl-2(1H)-pyridone, 2-Hydroxy-3-cyano-4-phenyl-6-(3-bromo-6-hydroxyphenyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H11BrN2O2
CAS Number:
Molecular Weight:
367.20
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Assay

≥90% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

This product has been discontinued.



A pyridone compound that acts as an ATP-competitive inhibitor of PIM1 kinase (IC50 = 50 nM) with excellent selectivity over PIM2 and MEK1/2 (IC50 >20 µM).

A pyridone compound that acts as an ATP-competitive inhibitor of PIM1 kinase (IC50 = 50 nM) with excellent selectivity over PIM2 and MEK1/2 (IC50 >20 µM).

Biochem/physiol Actions

Cell permeable: no
Primary Target
PIM1
Product competes with ATP.
Reversible: no
Target IC50: 50 nM against PIM1 kinase

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Cheney, I. W., et al. 2007. Bioorg. Med. Chem. Lett.17, 1679.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service