Skip to Content
Merck
All Photos(1)

Key Documents

524738

Sigma-Aldrich

Phosphodiesterase 5, Catalytic Domain, Human, Recombinant, S. frugiperda

Synonym(s):

PDE5CAT, cGMP-Specific Phosphodiesterase 5, Catalytic Domain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.54
Product 524738 is not currently sold in your country. Contact Technical Service

form

liquid

Quality Level

specific activity

≥0.5 U/μL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human, phosphodiesterase 5 catalytic subunit expressed in S. frugiperda insect cells using a baculovirus expression system. Supplied as a partially purified, catalytically active preparation.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmol cGMP to GMP per min at 30°C, pH 7.5.

Physical form

In 150 mM NaCl, 10 mM Tris-HCl, 0.1% NP-40, pH 8.3.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Gibson, L.C.D., et al. 2006. Eur. J. Pharmacol.538, 39.
Blount, M.A., et al. 2004. Mol. Pharmacol.66, 144.
Bakre, M.M., et al. 2000. J. Cell. Biochem.77, 159.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service