Skip to Content
Merck
All Photos(1)

Key Documents

524618

Sigma-Aldrich

PIP3 Antagonist, PITenin-7

The PIP3 Antagonist, PITenin-7 controls the biological activity of PIP3.

Synonym(s):

PIP3 Antagonist, PITenin-7, N-(4-Hydroxybiphenyl-3-ylcarbamothioyl)-3,5-dimethylbenzamide, Phosphatidylinositol-3,4,5-triphosphate Antagonist, PITenin-7, PIT-7, Akt Inhibitor XX, PDK1 Inhibitor V

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$203.00

$203.00


Estimated to ship on18 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$203.00

About This Item

Empirical Formula (Hill Notation):
C22H20N2O2S
CAS Number:
Molecular Weight:
376.47
UNSPSC Code:
12352200
NACRES:
NA.77

$203.00


Estimated to ship on18 April 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow-white

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable DM-PIT-1 (Cat. No. 524619) analog that is more selective and potent than DM-PIT-1 against PH domain-mediated PIP3 (Cat. No. 524615) binding to Akt1 (IC50 = 13.4 and 27.1 µM, respectively) and PDK1 (IC50 = 52.3 and 80.5 µM, respectively), while being much less active than DM-PIT-1 against PIP3 GRP1 PH domain binding (12% vs 84% inhibition with 100 µM respective compound). PIT-7 is also shown to be more effective than DM-PIT-1 in inhibiting human glioblastoma U87MG survival (IC50 = 6.6 vs. 33.9 µM, respectively) and cellular Akt Ser473 phosphorylation in cultures in vitro.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Miao, B., et al. 2010. Proc. Natl. Acad. Sci. USA107, 20126.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service