Skip to Content
Merck
All Photos(1)

Key Documents

516560

Sigma-Aldrich

Pertussis toxin

from Bordetella pertussis, Five distinct bands by SDS-PAGE, lyophilized, protein endotoxin, Calbiochem

Synonym(s):

Pertussis Toxin, Bordetella pertussis

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Product 516560 is not currently sold in your country. Contact Technical Service

Product Name

Pertussis Toxin, Bordetella pertussis, This Pertussis toxin, CAS 70323-44-3, catalyzes ADP-ribosylation of guanine nucleotide-binding regulatory proteins Gi, Go, and Gt.

description

RTECS - XW5883750

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

adenylate cyclase ≤35 pmol/min-μg (in the presence of calmodulin)

shipped in

ambient

storage temp.

2-8°C

SMILES string

O(C)C(=O)CCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C26H52O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-25-26(27)28-2/h3-25H2,1-2H3

InChI key

WOPKHAQDUMDJIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pertussis toxin (PT), secreted protein exotoxin and virulence factor[1] by Bordetella pertussis, is the causative agent of whooping cough.[2] It is transported across the bacterial outer membrane through a type IV secretion system encoded by the ptl genes, situated downstream from the ptx genes that encode the toxin. This complex multisubunit toxin with an AB5 configuration, serves as a crucial component of acellular pertussis vaccines. It can function protectively as a single-component toxoid vaccine. Moreover, it is widely used in mammalian cell signaling studies due to its inhibitory effect on G-protein-coupled receptor pathways. It inhibits neutrophil recruitment and airway influx, a common inflammatory response to bacterial infection.[1]

Application

Pertussis Toxin, Bordetella pertussis has been used as a Gi protein inhibitor to study its ability to reduce the depolarizing responses of hypothalamic pro-opiomelanocortin (POMC) neurons.[3] It has also been used in pertussis toxin experiments to block Gαi for an in vivo inner ear and an in vitro outer hair cell (HC) study.[4]

Biochem/physiol Actions

Primary Target
Guanine nucleotide-binding regulatory proteins Gi, Go, and Gt
Product does not compete with ATP.

Warning

Toxicity: Toxic (F)

Physical form

Lyophilized from 50 mM NaCl, 10 mM sodium phosphate buffer, pH 7.0.

Reconstitution

Following reconstitution, refrigerate (4°C). For long-term storage in solution, reconstitute with a high ionic strength buffer, such as 100 mM sodium phosphate, 500 mM sodium chloride, pH 7.0. Stock solutions are stable for at least 6 months for use in CHO cell assays and for 1-2 months for use in non-cellular systems.
Reconstitute with 500 µl H₂O. Obtain a uniform suspension before each use by scraping the walls of the tube and gently pipetting the solution to mix.

Analysis Note

Five distinct bands by SDS-PAGE

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Joshi, S.A., et al. 1998. FEBS Lett. 441, 67.
Fryer, M.W. 1992. Neurosci. Lett.146, 84.
Gierschik, P. 1992. Curr. Topics Microbiol. Immunol.175, 69.
Harada, J., et al. 1992. Eur. J. Pharmacol.227, 301.
Hausman, S.Z., and Burns, D.L. 1992. J. Biol. Chem.267, 13735.
Kaslow, H.R., and Burns, D.L., 1992. FASEB J.6, 2684.
Zhu, J., et al. 1992. J. Pharmacol. Exp. Therap.263, 1479.
Hewlett, E.L., et al. 1983. Infect. Immun.40, 1198.
Moss, J., et al. 1983. J. Biol. Chem.258, 11879.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pertussis Toxin: Structure-Function Relationship
Locht C, et al.
Bacterial Protein Toxins, 167-185 (2000)
Live imaging of hair bundle polarity acquisition demonstrates a critical timeline for transcription factor Emx2
Tona Y and Wu DK
eLife, 9, e59282-e59282 (2020)
Nicholas H Carbonetti
Future microbiology, 5(3), 455-469 (2010-03-10)
Pertussis toxin and adenylate cyclase toxin are two important virulence factors of Bordetella pertussis, the bacterial cause of the respiratory disease pertussis or whooping cough. In addition to studies on the structure, function and role in pathogenesis of these two
P Órdenes et al.
Scientific reports, 11(1), 21644-21644 (2021-11-06)
Previous studies indicate that the activity of hypothalamic POMC neurons can be regulated by glucose via intracellular mechanisms, but its regulation by lactate is poorly understood. In addition to its energetic role, lactate acts as a signaling molecule. In this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service