Skip to Content
Merck
All Photos(1)

Key Documents

512533

Sigma-Aldrich

MKLP-2 Inhibitor, Paprotrain

The MKLP-2 Inhibitor, Paprotrain controls the biological activity of MKLP-2. This small molecule/inhibitor is primarily used for Cell Signaling applications.

Synonym(s):

MKLP-2 Inhibitor, Paprotrain, (Z)-2-(1H-Indol-3-yl)-3-(pyridin-3-yl)acrylonitrile, PAssenger PROteins TRAnsport INhibitor, Mitotic Kinesin-Like Protein 2 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$307.00

$307.00


Estimated to ship on25 May 2025



Select a Size

Change View
25 MG
$307.00

About This Item

Empirical Formula (Hill Notation):
C16H11N3
Molecular Weight:
245.28
UNSPSC Code:
12352200
NACRES:
NA.77

$307.00


Estimated to ship on25 May 2025


Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

A cell-permeable acrylonitrile compound that inhibits the kinesin-6 family member MKLP-2 (Mitotic kinesin-like protein 2; IC50 against basal and microtubule-/MT-stimulated ATPase activity = 1.35 and 0.83 µM, respectively) in a reversible, ATP-uncompetitive (Ki = 3.36 µM), and MT-noncompetitive (Ki = 1.6 µM) manner, with little effect against the ATPase activity of 12 other kinesins (≤20% inhibition at 50 µM), including the close related MKLP-1 and MPP1 (M-phase phosphoprotein 1). Shown to inhibit cellular Aurora B and Survivin anaphase centromere-to-spindle midzone relocation in HeLa cultures (50 µM for 8 h), but not MKLP-1- or Kif4-mediated PRC1 relocation, resulting in misaligned chromosomes (70% of total metaphase cells), multipolar spindles (14% of total metaphase cells), and eventual apoptosis.

Packaging

Packaged under inert gas

Warning

Toxicity: Regulatory Review (Z)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tcherniuk, S., et al. 2010. Angew. Chem. Int. Ed.49, 8228.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Gulluni et al.
Science (New York, N.Y.), 374(6573), eabk0410-eabk0410 (2021-12-10)
Cytokinetic membrane abscission is a spatially and temporally regulated process that requires ESCRT (endosomal sorting complexes required for transport)–dependent control of membrane remodeling at the midbody, a subcellular organelle that defines the cleavage site. Alteration of ESCRT function can lead
Onur Sen et al.
Developmental cell, 56(22), 3082-3099 (2021-11-11)
Chromosome mis-segregation during mitosis leads to aneuploidy, which is a hallmark of cancer and linked to cancer genome evolution. Errors can manifest as "lagging chromosomes" in anaphase, although their mechanistic origins and likelihood of correction are incompletely understood. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service