Skip to Content
Merck
All Photos(1)

Key Documents

5.09880

Sigma-Aldrich

IDH2 Inhibitor, R140Q Somatic Mutant-Specific, AGI-6780

Synonym(s):

IDH2 Inhibitor, R140Q Somatic Mutant-Specific, AGI-6780, Isocitrate Dehydrogenase 2 R140 Somatic Mutant Inhibitor, AGI6780

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$279.00

$279.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
$279.00

About This Item

Empirical Formula (Hill Notation):
C21H18F3N3O3S2
CAS Number:
Molecular Weight:
481.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$279.00


Estimated to ship on31 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cc(ccc1)NC(=O)Nc2c(ccc(c2)[S](=O)(=O)NC4CC4)c3c[s]cc3

InChI

1S/C21H18F3N3O3S2/c22-21(23,24)14-2-1-3-16(10-14)25-20(28)26-19-11-17(32(29,30)27-15-4-5-15)6-7-18(19)13-8-9-31-12-13/h1-3,6-12,15,27H,4-5H2,(H2,25,26,28)

InChI key

CCAWRGNYALGPQH-UHFFFAOYSA-N

General description

A cell-permeable urea sulfonamide that inhibits the activity of Isocitrate Dehydrogenase 2 (IDH2) R140Q mutant-containing dimers in an α-ketoglutarate/α-KG-non-competitive and NADPH-uncompetitive manner by stabilizing the mutant dimers in an inactive conformation via a high-affinity allosteric interaction at the dimer interface with a slow-binding kinetic (kon = 5.8 x 104/M/min; koff = 8.3 x 10-3/min), while displaying much reduced potency against wt IDH2 dimer-catalyzed α-KG production and little or no activity toward wt IDH1 or R132H IDH2 mutant. Selectively inhibits 2-HG production in IDH2 R140Q, but not R132H, mutant-expressing U87 cultures (IC50 in 48 h = 11 nM vs. >100 µM). AGI-6780 treatment (5 µM) of primary bone marrow cultures from AML patients with R140Q mutation is shown to reactivate the differentiation of immature AML blasts without cytotoxicity.
A cell-permeable ureido-benzenesulfonamide that inhibits α-ketoglutarate (α-KG)-to-(R)-2-hydroxyglutarate (2-HG) conversion catalyzed by Isocitrate Dehydrogenase-2 (IDH2) R140Q mutant-containing dimers (IC50/preincubation time = 4 nM/16 h & 120 nM/1 h against R140Q-wt heterodimer; 23 nM/16 h & 170 nM/1 h against R140Q homodimer) in a substrate-non-competitive and NADPH-uncompetitive manner by targeting the dimer interface via a high affinity allosteric interaction with a slow-binding kinetic (kon = 5.8 x 104/M/min; koff = 8.3 x 10-3/min) and stabilizing the mutant dimers in an inactive conformation, while displaying much reduced potency against wt IDH2 dimer-catalyzed α-KG production (IC50/preincubation time = 190 nM/16 h & 2.7 µM/1 h) and little or no activity toward LDHA, 3PGDH, GDH, G6PDH, R132H mutant or wt IDH1. Selectively inhibits 2-HG production in R140Q IDH2-expressing U87 glioblastoma & TF-1 erythroleukemia, but not R132H-expressing U87 cultures (IC50 in 48 h = 11 nM, 18 nM, and >100 µM, respectively) and sensitizes R140Q IDH2-expressing TF-1 to EPO-induced erythroid differentiation. Likewise, inhibition of 2-HG production in primary bone marrow cultures from AML patients with R140Q mutation (complete inhibition with 5 µM AGI-6780) is shown to reactivate the differentiation of immature AML blasts without cytotoxicity. IDH1 Inhibitor, R132 Somatic Mutant-Specific, AGI-5198 (>Cat. No. 410972) is also available.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
(IDH2) R140Q mutants
Reversible: yes
Target IC50: 4 nM/16 h & 120 nM/1 h against R140Q-wt heterodimer

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Wang, F., et al. 2013. Science340, 622.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service