Skip to Content
Merck
All Photos(2)

Key Documents

5.08605

Sigma-Aldrich

Sodium Orthovanadate, Activated

Synonym(s):

Sodium Orthovanadate, Activated, Orthovanadate, Activated, Vanadate, Activated

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
$206.00
5 ML
$206.00

$206.00


Estimated to ship on01 June 2025



Select a Size

Change View
1 SET
$206.00
5 ML
$206.00

About This Item

Empirical Formula (Hill Notation):
Na3O4V
Molecular Weight:
183.91
UNSPSC Code:
12352202
NACRES:
NB.21

$206.00


Estimated to ship on01 June 2025


form

liquid

Quality Level

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
protect from light

color

colorless

storage temp.

−70°C

General description

A depolymerized vanadate prepared from Sodium Orthovanadate (>Cat. No. 567540) that exhibits significant enhanced ability to inhibit endogenous phosphatases in a competitive manner. Preserves the phosphorylation state in cells and cell lysates. Completely reversible with EDTA and dilution. Recommended concentration in lysis buffer ~ 1 mM. Available as 5 ml or as a set of 3 x 1 ml.

Biochem/physiol Actions

Primary Target
Phosphatases
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic (F)

Physical form

Supplied as 10 mg/ml in aqueous NaOH, pH 10.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 1 year at -70°C.

Other Notes

Gordon, J.A. 1984. Methods. Enzymol.201, 477.
Brown, D.J., and Gordon, J.A. 1984. J. Biol. Chem.259, 9580.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Did the use of 508605 Sodium Orthovanadate at 1 mM in whole blood result in cell death?

    1 answer
    1. The recommended working concentration for cell lysate application is 1 mM. It's important to note that orthovanadate can be toxic in the blood at a concentration of 0.12 mM. For further information, you can refer to the publication "Biochim Biophys Acta. 2012 Sep;1818(9):2260-70" with the PMID: 22546530. This publication discusses the effects of sodium orthovanadate on human erythrocytes and neuroblastoma cells in vitro. It mentions that the lethal concentration of vanadium in the blood of a patient was calculated to be about 0.12 mM.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service